Skip to Content
Merck
All Photos(6)

Key Documents

HPA005857

Sigma-Aldrich

Anti-FBP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-D-fructose-1,6- bisphosphate 1-phosphohydrolase 1, Anti-FBPase 1, Anti-Fructose-1,6-bisphosphatase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

DCLVSVGTIFGIYRKKSTDEPSEKDALQPGRNLVAAGYALYGSATMLVLAMDCGVNCFMLDPAIGEFILVDKDVKIKKKGKIYSLNEGYAKDFDPAVTEYIQRKKFPPDNSAP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FBP1(2203)

General description

Fructose-1,6-bisphosphatase 1, is a homotetrameric protein, encoded by the gene mapped to human chromosome 9q22.

Immunogen

Fructose-1,6-bisphosphatase 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-FBP1 antibody produced in rabbit has been used in immunohistochemical staining and immunoblotting.

Biochem/physiol Actions

Fructose-1,6-bisphosphatase 1 (FBP1) is a rate-limiting enzyme in gluconeogenesis. It catalyzes the hydrolysis of fructose-1,6-bisphosphate (F-1,6-BP) to fructose 6-phosphate and inorganic phosphate. FBP is a potential therapeutic target in the treatment of type II diabetes. It plays a major role in glucose metabolism in cancers. In addition, it also regulates metabolism of breast cancer cells by inhibiting hypoxia-inducible factor 1α (HIF-1α).
Mutation in the FBP1 gene is associated with clear cell renal cell carcinoma (ccRCC). Deficiency of this protein is an autosomal, recessively inherited disease that progresses with severe hypoglycemia and metabolic attacks result in a defect in gluconeogenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70322

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Safia Akhtar et al.
Scientific reports, 11(1), 11367-11367 (2021-06-02)
Recent studies suggested that renal gluconeogenesis is substantially stimulated in the kidney in presence of obesity. However, the mechanisms responsible for such stimulation are not well understood. Recently, our laboratory demonstrated that mice fed high fat diet (HFD) exhibited increase
Juan Yang et al.
Carcinogenesis, 38(2), 134-143 (2016-11-01)
Reprogrammed metabolism has been identified as an emerging hallmark in cancer cells. It has been demonstrated that fructose-1, 6-bisphosphatase 1 (FBP1) as a rate-limiting enzyme in gluconeogenesis plays critical roles in tumor initiation and progression in several cancer types. However
Loss of FBP1 facilitates aggressive features of hepatocellular carcinoma cells through the Warburg effect
Yang J, et al.
Carcinogenesis, 38(2) (2017)
FBP1 modulates cell metabolism of breast cancer cells by inhibiting the expression of HIF-1alpha
Shi L, et al.
Neoplasma, 64(4), 535-542 (2017)
Bushra Afroze et al.
European journal of pediatrics, 172(9), 1249-1253 (2013-07-25)
Fructose-1,6-bisphosphatase (FBP) deficiency is an autosomal-recessive disorder of gluconeogenesis resulting from mutations within the FBP1 gene. During periods of trivial illness, individuals with FBP deficiency may develop ketotic hypoglycemia, metabolic acidosis, lactic acidemia, and an increased anion gap. Although detection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service