Skip to Content
Merck
All Photos(9)

Key Documents

HPA005679

Sigma-Aldrich

Anti-HPGD antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-15-Hydroxyprostaglandin dehydrogenase [NAD+] antibody produced in rabbit, Anti-PGDH antibody produced in rabbit, Anti-Prostaglandin dehydrogenase 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

LAANLMNSGVRLNAICPGFVNTAILESIEKEENMGQYIEYKDHIKDMIKYYGI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HPGD(3248)

Looking for similar products? Visit Product Comparison Guide

General description

15-hydroxyprostaglandin dehydrogenase (HPGD) is a prostaglandin-degrading enzyme and belongs to the short chain dehydrogenases/reductase family. The gene HPGD is mapped to human chromosome 4.

Immunogen

15-Hydroxyprostaglandin dehydrogenase [NAD+] recombinant protein epitope signature tag (PrEST)

Application

Anti-HPGD antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

15-hydroxyprostaglandin dehydrogenase (HPGD) has a functional colon cancer tumor suppressor activity. This enzyme catalyzes the inactivating NAD+-dependent conversion of the prostaglandin 15-OH group to a 15-keto group. Its expression is directly controlled and strongly induced by activation of the TGF-β tumor suppressor pathway. The enzymatic pathway that induces colon cancer suppression is activated by TGF-β and mediated by HPGD. It also catalyzes the NAD-dependent dehydrogenation of lipoxin A4 (LXA4) to form 15-oxo-lipoxin A4 by the dehydrogenation of the C15 hydroxyl group of LXA4 to an oxo- group hence forming 15-oxo-LXA4. Mutations in this gene can cause primary hypertrophic osteoarthropathy and cranioosteoarthropathy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70117

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas J Monteleone et al.
Scientific reports, 9(1), 5405-5405 (2019-04-02)
Elevated prostaglandin E2 (PGE2) levels are observed in colorectal cancer (CRC) patients, and this increase is associated with poor prognosis. Increased synthesis of PGE2 in CRC has been shown to occur through COX-2-dependent mechanisms; however, loss of the PGE2-catabolizing enzyme
C B Clish et al.
The Journal of biological chemistry, 275(33), 25372-25380 (2000-06-06)
The lipoxins (LX) are autacoids that act within a local inflammatory milieu to dampen neutrophil recruitment and promote resolution. 15-Hydroxyprostaglandin dehydrogenase (15-PGDH) and 15-oxoprostaglandin 13-reductase, also termed leukotriene B(4) 12-hydroxydehydrogenase (PGR/LTB(4)DH), are two enzymatic activities appreciated for their roles in
Wenke Seifert et al.
European journal of human genetics : EJHG, 17(12), 1570-1576 (2009-07-02)
Cranio-osteoarthropathy, clinically classified as a variant of primary hypertrophic osteoarthropathy, is a very rare autosomal-recessive condition characterized by delayed closure of the cranial sutures and fontanels, digital clubbing, arthropathy, and periostosis. Recently, mutations in the gene HPGD, which encodes the
Carsten Bergmann et al.
Experimental dermatology, 20(6), 531-533 (2011-03-24)
Digital clubbing is the most prominent feature in primary (PHO) and secondary (pulmonary) hypertrophic osteoarthropathy (HO). Homozygous and compound heterozygous germline mutations in the 15-hydroxyprostaglandin dehydrogenase (HPGD) gene encoding the major prostaglandin PGE2 catabolizing enzyme have been recently described in
M Tariq et al.
Journal of medical genetics, 46(1), 14-20 (2008-09-23)
Isolated congenital nail clubbing (ICNC) is a rare autosomal recessive disorder characterised by enlargement of the terminal segments of fingers and toes with thickened nails due to proliferation of the connective tissues and abnormal function of the nail matrix. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service