Skip to Content
Merck
All Photos(7)

Documents

HPA003868

Sigma-Aldrich

Anti-MIF antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GIF antibody produced in rabbit, Anti-Glycosylation-inhibiting factor antibody produced in rabbit, Anti-Macrophage migration inhibitory factor antibody produced in rabbit, Anti-Phenylpyruvate tautomerase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:2500-1:5000

immunogen sequence

FIVNTNVPRASVPDGFLSELTQQLAQATGKPPQYIAVHVVPDQLMAFGGSSEPCALCSLHSIGKIGGAQNRSYSKLLCGLLAERLRISPDRVYINYYDMNAANVGWNN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

glycosylation

Gene Information

human ... MIF(4282)

Immunogen

Macrophage migration inhibitory factor recombinant protein epitope signature tag (PrEST)

Application

Anti-MIF antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Macrophage migration inhibitory factor (MIF, MMIF and GIF) is a protein encoded by the MIF gene in humans and is mapped to chromosome 10. It is a potent cytokine that plays an important role in the regulation of immune responses. It acts as an integral component of the host antimicrobial alarm system and stress response that promotes the pro-inflammatory functions of immune cells. It is implicated in the pathogenesis of sepsis, and inflammatory and autoimmune diseases. It may also be implicated in both types of diabetes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73599

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuriko I Sánchez-Zamora et al.
Journal of diabetes research, 2014, 804519-804519 (2014-02-15)
Autoimmunity and chronic low-grade inflammation are hallmarks of diabetes mellitus type one (T1DM) and type two (T2DM), respectively. Both processes are orchestrated by inflammatory cytokines, including the macrophage migration inhibitory factor (MIF). To date, MIF has been implicated in both
Johannes Dominikus Pallua et al.
Journal of proteomics, 91, 500-514 (2013-08-21)
New biomarkers are needed to improve the specificity of prostate cancer detection and characterisation of individual tumors. In a proteomics profiling approach using MALDI-MS tissue imaging on frozen tissue sections, we identified discriminating masses. Imaging analysis of cancer, non-malignant benign
Yu-Huei Liu et al.
PloS one, 9(3), e92849-e92849 (2014-03-29)
Graves disease (GD) is an autoimmune disease. Macrophage migration inhibitory factor (MIF) is a potent cytokine that plays an important role in the regulation of immune responses. Two polymorphisms in the promoter region of MIF, rs5844572 and rs755622, are known
U De la Cruz-Mosso et al.
Human immunology, 75(5), 433-439 (2014-02-18)
Macrophage migration inhibitory factor (MIF) is an upstream immunoregulatory cytokine associated with the pathogenesis of autoimmune inflammatory diseases. There is evidence that MIF functions in a positive feedback loop with TNF-α that could perpetuate the inflammatory process in systemic lupus
Jelena Põlajeva et al.
Molecular oncology, 8(1), 50-58 (2013-10-05)
Recently, glioma research has increased its focus on the diverse types of cells present in brain tumors. We observed previously that gliomas are associated with a profound accumulation of mast cells (MCs) and here we investigate the underlying mechanism. Gliomas

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service