Skip to Content
Merck
All Photos(5)

Key Documents

HPA003548

Sigma-Aldrich

Anti-Sept3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Neuronal-specific septin-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

LSKVVNIIPVIAKADTMTLEEKSEFKQRVRKELEVNGIEFYPQKEFDEDLEDKTENDKIRQESMPFAVVGSDKEYQVNGKRVLGRKTPWGIIEVENLNHCEFALLRDFVIRTHLQDLKEVTHNIHYETYRAKRLNDNGG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SEPT3(55964)

Immunogen

Neuronal-specific septin-3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Neuronal-specific septin-3 is a protein encoded by the SEPT3 gene in humans. It is a novel member of the septin subfamily of GTPase domain proteins in human neuronal cells. It is a neuron-specific protein highly enriched in nerve terminals and supports a secretory role in synaptic vesicle recycling. The protein is involved in vesicle trafficking, neurite outgrowth and neurofibrillary tangle formation. It is phosphorylated by cGMP-dependent protein kinase I (PKG-I) and the cGMP/PKG pathway and is involved in presynaptic plasticity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73554

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Xue et al.
Journal of neurochemistry, 91(3), 579-590 (2004-10-16)
The septins are GTPase enzymes with multiple roles in cytokinesis, cell polarity or exocytosis. The proteins from the mammalian septin genes are called Sept1-10. Most are expressed in multiple tissues, but the mRNA for Sept5 (CDCrel-1) and Sept3 (G-septin) appear
Masanori Takehashi et al.
Gene expression, 11(5-6), 263-270 (2004-06-18)
Septin 3 is a novel member of the septin subfamily of GTPase domain proteins that was recently identified in human neuronal cells. These proteins are involved in vesicle trafficking, neurite outgrowth, and neurofibrillary tangle formation; however, the expression and functional
Ramona Miske et al.
Journal of neuroinflammation, 20(1), 88-88 (2023-03-31)
Septins are cytoskeletal proteins with filament forming capabilities, which have multiple roles during cell division, cellular polarization, morphogenesis, and membrane trafficking. Autoantibodies against septin-5 are associated with non-paraneoplastic cerebellar ataxia, and autoantibodies against septin-7 with encephalopathy with prominent neuropsychiatric features.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service