Skip to Content
Merck
All Photos(5)

Key Documents

HPA003512

Sigma-Aldrich

Anti-RPLP0 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-60S acidic ribosomal protein P0 antibody produced in rabbit, Anti-L10E antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

VVLMGKNTMMRKAIRGHLENNPALEKLLPHIRGNVGFVFTKEDLTEIRDMLLANKVPAAARAGAIAPCEVTVPAQNTGLGPEKTSFFQALGIT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPLP0(6175)

Immunogen

60S acidic ribosomal protein P0 recombinant protein epitope signature tag (PrEST)

Application

Anti-RPLP0 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

60S acidic ribosomal protein P0 is a protein encoded by the RPLP0 gene in humans and is found in active polysomes as well as in the nucleolus. The protein is an essential component of the eukaryotic ribosomal stalk and interacts with the helix-loop-helix protein human Grap2 and cyclin D interacting protein (GCIP)/D-type cyclin-interacting protein 1/human homolog of MAID protein. The overexpression of this gene may lead to tumor-genesis in breast and liver tissues.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86617

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Repression of Activated Aryl Hydrocarbon Receptor?Induced Transcriptional Activation by 5a-Dihydrotestosterone in Human Prostate Cancer LNCaP and Human Breast Cancer T47D Cells
Sanada N, et al.
Journal of Pharmaceutical Sciences, 109, 380-387 (2009)
Noriko Sanada et al.
Journal of pharmacological sciences, 109(3), 380-387 (2009-03-10)
Polycyclic aromatic hydrocarbons (PAHs) and dioxins are ubiquitous environmental pollutants and activate the aryl hydrocarbon receptor (AhR), a ligand-activated transcription factor. It has been reported that testosterone represses 2,3,7,8-tetrachlorodibenzo-p-dioxin-induced transcription of the cytochrome P450 (CYP) 1A1 gene in LNCaP cells.
Kathrin Guitart et al.
Glia, 64(6), 896-910 (2016-03-19)
Prion protein (PrP) protects neural cells against oxidative stress, hypoxia, ischemia, and hypoglycemia. In the present study we confirm that cultured PrP-deficient neurons are more sensitive to oxidative stress than wild-type neurons and present the novel findings that wild-type, but
H Towbin et al.
The Journal of biological chemistry, 257(21), 12709-12715 (1982-11-10)
Mice were immunized against chick ribosomes with the use of various protocols and immunogen preparations. Hybridomas were prepared, clones screened, and specific antibodies identified by reversible protein staining followed by immunoperoxidase staining on nitrocellulose blots. Clones were obtained which secreted
T-W Chang et al.
Oncogene, 27(3), 332-338 (2007-07-11)
The ribosomal acidic P0 protein, an essential component of the eukaryotic ribosomal stalk, was found to interact with the helix-loop-helix protein human Grap2 and cyclin D interacting protein (GCIP)/D-type cyclin-interacting protein 1/human homolog of MAID protein. Using in vivo and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service