Skip to Content
Merck
All Photos(4)

Key Documents

HPA001862

Sigma-Aldrich

ANTI-IRF9 antibody produced in rabbit

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-IFN-α-responsive transcription factor subunit, Anti-IRF9, Anti-ISGF-3 γ, Anti-ISGF-3γ, Anti-ISGF3 p48 subunit, Anti-ISGF3G, Anti-Interferon regulatory factor 9, Anti-Interferon-stimulated gene factor 3 γ, Anti-Transcriptional regulator ISGF3 subunit gamma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

EEEDAMQNCTLSPSVLQDSLNNEEEGASGGAVHSDIGSSSSSSSPEPQEVTDTTEAPFQGDQRSLEFLLPPEPDYSLLLTFIYNGRVVGEAQVQSLDCRLVAEPSGSESSMEQVLFPKPGPLEPTQRLLSQLER

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IRF9(10379)

Specificity

Note: The Ensemble Gene ID has changed from ENSG00000092098 in the 46:36 release of the database to ENSG00000213928 in the 48:36 release of the database.

Immunogen

Interferon regulatory factor 9 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

STAT-containing transcription factor, ISGF3 (interferon-stimulated gene factor 3) has two independent components, termed ISGF3α, regulatory component for interferon signaling, and ISGF3γ, the DNA recognition subunit. It positively regulates the initial activation of Interferons-α-responsive genes. Activated α subunit of ISGF3 helps to regulate the ISGF3γ-DNA interactions as well as stabilize the ternary complex. In nucleus, ISGF3γ identifies IFN (interferon)-stimulated response element (ISRE) recognition component of the transcription factor and shows ISRE-binding specificity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77486

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D S Kessler et al.
Genes & development, 4(10), 1753-1765 (1990-10-01)
The interaction of interferon-alpha (IFN-alpha) with a specific cell-surface receptor elicits physiological changes that rely on rapid transcriptional activation of a group of IFN-alpha-stimulated genes (ISGs). The IFN-stimulated response element (ISRE), a conserved regulatory element of all ISGs, is the
Lili Liao et al.
eLife, 7 (2018-10-26)
Whereas VHL inactivation is a primary event in clear cell renal cell carcinoma (ccRCC), the precise mechanism(s) of how this interacts with the secondary mutations in tumor suppressor genes, including PBRM1, KDM5C/JARID1C, SETD2, and/or BAP1, remains unclear. Gene expression analyses
S A Veals et al.
Molecular and cellular biology, 12(8), 3315-3324 (1992-08-11)
Alpha interferon stimulates transcription by converting the positive transcriptional regulator ISGF3 from a latent to an active form. This receptor-mediated event occurs in the cytoplasm, with subsequent translocation of the activated factor to the nucleus. ISGF3 has two components, termed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service