Skip to Content
Merck
All Photos(6)

Key Documents

HPA001667

Sigma-Aldrich

Anti-PAPPA antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-IGF-dependent IGFBP-4 protease, Anti-IGFBP-4ase, Anti-Insulin-like growth factor-dependent IGF-binding protein 4 protease, Anti-PAPP-A, Anti-Pappalysin-1, Anti-Pregnancy-associated plasma protein A

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

SCLDHNSESIILPMNVTVRDIPHWLNPTRVERVVCTAGLKWYPHPALIHCVKGCEPFMGDNYCDAINNRAFCNYDGGDCCTSTVKTKKVTPFPMSCDLQGDCACRDPQAQEHSRKDL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PAPPA(5069)

Immunogen

Pappalysin-1 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Pregnancy-associated plasma protein-A (PAPPA) is an insulin-like growth factor binding protein (IGFBP) protease. It is characterized as a zinc-binding matrix metalloproteinase that is abundantly expressed in eroded and ruptured plaques. It may function as a marker of plaque destabilization. In presence of IGF, PAPP-A particularly cleaves IGF-binding protein (IGFBP)-4, one of six antagonists of IGF action, which leads to the release of IGF bound to IGFBP-4.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77476

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Kirschner et al.
Oncoimmunology, 6(2), e1273301-e1273301 (2017-03-28)
Pregnancy-associated plasma protein-A (PAPPA), also known as pappalysin, is a member of the insulin-like growth factor (IGF) family. PAPPA acts as a protease, cleaving IGF inhibitors, i.e., IGF binding proteins (IGFBPs), thereby setting free IGFs. The insulin/IGF-axis is involved in
Christopher Heeschen et al.
Journal of the American College of Cardiology, 45(2), 229-237 (2005-01-18)
The goal of this study was to determine the predictive value of pregnancy-associated plasma protein-A (PAPP-A) in patients with acute coronary syndromes (ACS). Pregnancy-associated plasma protein-A is a zinc-binding matrix metalloproteinase abundantly expressed in eroded and ruptured plaques and may
M T Overgaard et al.
The Journal of biological chemistry, 275(40), 31128-31133 (2000-07-27)
Pregnancy-associated plasma protein-A (PAPP-A), originally known from human pregnancy serum, has recently been demonstrated to be a metzincin superfamily metalloproteinase involved in normal and pathological insulin-like growth factor (IGF) physiology. PAPP-A specifically cleaves IGF-binding protein (IGFBP)-4, one of six antagonists
C A Conover et al.
Endocrinology, 142(5), 2155-2155 (2001-04-24)
Insulin-like growth factors (IGFs), IGF binding proteins (IGFBPs), and IGFBP proteases are important in ovarian function. IGFs stimulate granulosa steroidogenesis, an effect that is inhibited by IGFBP-4 and augmented by IGFBP-4 proteolysis. We have recently identified the IGFBP-4 protease in
Shi-Yuan Fan et al.
International journal of nanomedicine, 15, 2085-2094 (2020-04-11)
Pregnancy-associated plasma protein-A and -A2 (PAPP-A and -A2) are principally expressed in placental trophoblasts and play a critical role in the regulation of fetal and placental growth. PAPP-A2 shares 45% amino acid similarity with PAPP-A. This study aimed to investigate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service