Skip to Content
Merck
All Photos(8)

Key Documents

HPA001654

Sigma-Aldrich

Anti-APOH antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-β-2-Glycoprotein I antibody produced in rabbit, Anti-β2-GPI antibody produced in rabbit, Anti-β2-Glycoprotein 1 precursor antibody produced in rabbit, Anti-β2-Glycoprotein I antibody produced in rabbit, Anti-APC inhibitor antibody produced in rabbit, Anti-Activated protein C-binding protein antibody produced in rabbit, Anti-Anticardiolipin cofactor antibody produced in rabbit, Anti-Apo-H antibody produced in rabbit, Anti-Apolipoprotein H antibody produced in rabbit, Anti-B2GPI antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
independent
RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

ECREVKCPFPSRPDNGFVNYPAKPTLYYKDKATFGCHDGYSLDGPEEIECTKLGNWSAMPSCKASCKVPVKKATVVYQGERVKIQEKFKNGMLHGDKVSFFCKNKEKKCSYTEDAQCIDGTIEVPKCFKEHSSLAFSKTDASDVK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APOH(350)

Looking for similar products? Visit Product Comparison Guide

General description

APOH (apolipoprotein H, β-2-glycoprotein I) is a single chain polymorphic glycoprotein involved in phospholipid binding interactions on platelets. It exists in plasma both in a free form and in combination with lipoprotein particles. It contains 11 disulfide bridges and four N-linked glycosylation sites with molecular mass of ~43kDa. It comprises of four short consensus repeat (SCRs) from the complement control protein (CCP) module type and an extra fifth C-terminal domain.

Immunogen

β2-Glycoprotein 1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

APOH (apolipoprotein H, β-2-glycoprotein I) is associated with a variety of physiological pathways, including blood coagulation and immune response. It plays an essential role as a cofactor for the binding of certain antiphospholipid antibodies (APA) to anionic phospholipid. It consists of a positively charged lysine-rich domain which binds to negatively charged surfaces including heparin, DNA, cell membranes, endothelial cells,macrophages and acidic phospholipids.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84509

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M I Kamboh et al.
Lupus, 7 Suppl 2, S10-S13 (1998-11-14)
Apolipoprotein H (apoH; also known as beta2-glycoprotein I), is an essential cofactor for the binding of certain antiphospholipid antibodies (APA) to anionic phospholipid. The gene coding for apoH is polymorphic, with the occurrence of several common alleles in the general
Ruitong Gao et al.
The Journal of rheumatology, 43(11), 2026-2032 (2016-11-03)
Antiphospholipid-associated nephropathy (aPLN) is a severe condition in patients with lupus nephritis (LN). aPLN should be distinguished from other reasons for renal ischemia. The most important cofactor of antiphospholipid antibodies (aPL), β2-glycoprotein I (β2GPI), was shown in vitro to bind
H Mehdi et al.
Human genetics, 105(1-2), 63-71 (1999-09-10)
Apolipoprotein H (apoH, protein; APOH, gene) is a single chain glycoprotein that exists in plasma both in a free form and in combination with lipoprotein particles. ApoH has been implicated in several physiologic pathways, including lipid metabolism, coagulation, and the
Yi-Chen Lee et al.
Anticancer research, 43(8), 3455-3462 (2023-07-28)
Beta2-glycoprotein I (β2-GPI) is a plasma glycoprotein, which has been implicated in a variety of physiological functions. However, the connection between β2-GPI and breast cancer is mostly unknown. Breast cancer is a malignant tumor that severely impairs women's health worldwide.
Guiting Zhang et al.
Molecular medicine reports, 23(2) (2020-12-24)
Patients with antiphospholipid syndrome have been identified to have higher incidence rates of atherosclerosis (AS) due to the elevated levels of anti‑β2‑glycoprotein I (β2GPI) antibody (Ab). Our previous studies revealed that the anti‑β2GPI Ab formed a stable oxidized low‑density lipoprotein (oxLDL)/β2GPI/anti‑β2GPI Ab complex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service