Skip to Content
Merck
All Photos(6)

Key Documents

HPA001609

Sigma-Aldrich

Anti-HNRNPA1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HNRPA1, Anti-heterogeneous nuclear ribonucleoprotein A1, Anti-hnRNP-A1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

ETTDESLRSHFERWGMLTDCAVMRDPNTKRSRGFGFVTYATVEEVDAATNARPHKVDGKVVEPRRTVSREDYQRSGAHLTVKKIFVGGIKENTEKHQLRDYFEQHGKMEVIEIMTEAVARKGALPL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HNRNPA1(3178)

General description

HNRNPA1 (heterogeneous nuclear ribonucleoprotein A1) is a ribonucleoprotein, which functions as an RNA-binding protein. It can interact with both RNA and DNA, through its two nucleic acid-binding domains. It is proteolytically cleaved into a 196-amino acid protein called UP1 (unwinding protein 1). This protein retains the two parent nucleic acid-binding domains. It is one of the major RNA-binding proteins, and though it localizes to nucleus, it can shuttle to cytoplasm as well.

Immunogen

heterogeneous nuclear ribonucleoprotein A1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

HNRNPA1 (heterogeneous nuclear ribonucleoprotein A1) functions in mRNA metabolism. It shuttles continuously between nucleus and cytoplasm and functions in several processes such as mRNA export, internal ribosome entry site (IRES)-mediated translation and mRNA stability. In humans, it acts as an auxiliary factor for the processing of a specific miRNA substrate by binding specifically to pri-miR-18a, the stem-loop precursor of miR-18a and regulating its Drosha-mediated processing. Defects in this gene are associated with multisystem proteinopathy and ALS (Amyotrophic lateral sclerosis 20). An alternatively spliced isoform of this protein is capable of controlling the splicing and replication of human immunodeficiency virus (HIV)-1.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74297

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacques Jean-Philippe et al.
Biochimica et biophysica acta, 1839(4), 251-258 (2014-02-18)
Heterogeneous nuclear ribonucleoprotein A1 (hnRNP A1) is one of the most abundant RNA binding proteins. hnRNP A1 is localized prevalently in the nucleus but it can relocate to the cytoplasm in response to specific stimuli shuttling between nuclear and cytoplasmic
Xiaotong Gao et al.
Theranostics, 9(19), 5642-5656 (2019-09-20)
Rationale: Reciprocal interactions between leukemic cells and bone marrow mesenchymal stromal cells (BMMSC) remodel the normal niche into a malignant niche, leading to leukemia progression. Exosomes have emerged as an essential mediator of cell-cell communication. Whether leukemic exosomes involved in
Jason S Hudson et al.
Biochemistry, 53(20), 3347-3356 (2014-05-17)
The specific recognition by proteins of G-quadruplex structures provides evidence of a functional role for in vivo G-quadruplex structures. As previously reported, the ribonucleoprotein, hnRNP Al, and it is proteolytic derivative, unwinding protein 1 (UP1), bind to and destabilize G-quadruplex
Rui-Sheng Ke et al.
International journal of biological macromolecules, 129, 140-151 (2019-02-08)
Previous study has shown heterogeneous nuclear ribonucleoprotein A1(HNRNPA1) is highly expressed in various human cancers. In order to study the clinical value and potential function of HNRNPA1 in HBV-related hepatocellular carcinoma (HCC), three datasets from the GEPIA, GEO and TCGA
Sonia Guil et al.
Nature structural & molecular biology, 14(7), 591-596 (2007-06-15)
hnRNP A1 is an RNA-binding protein involved in various aspects of RNA processing. Use of an in vivo cross-linking and immunoprecipitation protocol to find hnRNP A1 RNA targets resulted in the identification of a microRNA (miRNA) precursor, pre-miR-18a. This microRNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service