Skip to Content
Merck
All Photos(4)

Key Documents

HPA001378

Sigma-Aldrich

Anti-PNN antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-140 kDa nuclear and cell adhesion-related phosphoprotein antibody produced in rabbit, Anti-DRS-protein antibody produced in rabbit, Anti-DRSP antibody produced in rabbit, Anti-Desmosome-associated protein antibody produced in rabbit, Anti-Domain-rich serine protein antibody produced in rabbit, Anti-Melanoma metastasis clone A protein antibody produced in rabbit, Anti-Nuclear protein SDK3 antibody produced in rabbit, Anti-Pinin antibody produced in rabbit, Anti-SR-like protein antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

EQKVELAQLQEEWNEHNAKIIKYIRTKTKPHLFYIPGRMCPATQKLIEESQRKMNALFEGRRIEFAEQINKMEARPRRQSMKEKEHQVVRNEEQKAEQEEGKVAQREEELEETGNQHNDVEIEEAGEEEEKEIAIVHSDAEKE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PNN(5411)

Immunogen

Pinin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PNN (pinin, desmosome associated protein) is a 140kDa phosphoprotein which is present in desmosomes and nuclear speckles. It colocalizes with mRNA splicing factors in nuclear speckled domains and plays an important role in mRNA processing. It is involved in the regulation of cell-cell adhesion of corneal epithelial cells. It also interacts with transcriptional repressor and pre-mRNA processing machinery in the nucleus. It is associated with the desmosome-intermediate filament complex and may be involved in pinning the intermediate filaments to the desmosome. Pinin is also involved in the regulation of the activity of several tumor suppressor genes. Due to its dual localization, pinin acts as a moonlighting protein with more than one function.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70425

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiang-Yu Meng et al.
American journal of cancer research, 11(4), 1286-1303 (2021-05-06)
Pinin (PNN), a desmosome associated protein, was demonstrated to be over-expressed and act as a tumor-promoting factor in ovarian cancer, hepatocellular carcinoma and colorectal cancer. However, the precise role of PNN in prostate cancer is still unknown. In the study
Chin Li et al.
Molecular and cellular biology, 23(20), 7363-7376 (2003-10-01)
Pnn/DRS protein is associated with desmosomes and colocalizes with splicing factors in nuclear speckled domains. The potential interaction of Pnn with RNPS1, a pre-mRNA splicing factor and a component of the exon-exon junction complex, prompted us to examine whether Pnn
Ping Wang et al.
Biochemical and biophysical research communications, 294(2), 448-455 (2002-06-08)
Pre-mRNA splicing occurs in a large macromolecular RNA-protein complex called the spliceosome. The major components of the spliceosome include snRNP and SR proteins. We have previously identified an SR-like protein, pinin (pnn), which is localized not only in nuclear speckles
Y Shi et al.
Oncogene, 19(2), 289-297 (2000-01-25)
Several cell adhesion-related proteins have been shown to act as tumor-suppressors (TS) in the neoplastic progression of epithelial-derived tumors. Pinin/DRS/memA was first identified in our laboratory and it was shown to be a cell adhesion-related molecule. Our previous study demonstrated
P Ouyang et al.
The Journal of cell biology, 135(4), 1027-1042 (1996-11-01)
We have identified a protein named pinin that is associated with the mature desmosomes of the epithelia (Ouyang, P., and S.P. Sugrue. 1992. J. Cell Biol. 118:1477-1488). We suggest that the function of pinin is to pin intermediate filaments to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service