Skip to Content
Merck
All Photos(7)

Key Documents

HPA001238

Sigma-Aldrich

Anti-MMP9 antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

CLG4B

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP9(4318)

General description

Matrix metalloproteinase-9 (MMP-9) is a pro-inflammatory, zinc-dependent proteinase, which is also known as 92kDa gelatinase. It is secreted by granulocytes and mononuclear cells. The activity of MMP-9 is controlled by gene transcription. The gene encoding it is localized on human chromosome 20q11.2-q13.1 and has 13 exons.

Immunogen

Recombinant protein corresponding to Matrix metalloproteinase-9

Sequence
PRQRQSTLVLFPGDLRTNLTDRQLAEEYLYRYGYTRVAEMRGESKSLGPALLLLQKQLSLPETGELDSATLKAMRTPRCGVPDLGRFQTFEGDLKWHHHNITYWIQNYSEDLPRAVIDDAFARAFALWSAVTPLTFTRVYSRDADI

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-MMP9 antibody produced in rabbit has been used in:
  • immunohistochemistry
  • western blotting
  • fluorescence immunohistochemistry

Biochem/physiol Actions

Matrix metalloproteinase-9 (MMP-9) catalyzes the degradation of gelatin and collagen. It has been studied as a biomarker for inflammation in multiple sclerosis (MS). The MMP9 protein is a zinc-dependent protease which is responsible for degradation of extracellular matrix components. It also participates in the pathogenesis of cancers.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83039

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peixin Dong et al.
Scientific reports, 6, 35480-35480 (2016-10-22)
Derepression of wild-type p53 by suppressing its negative inhibitor iASPP (Inhibitor of apoptosis-stimulating protein of p53) represents a potential therapeutic option for cervical cancer (CC). Here, we reported a novel functional significance of iASPP upregulation in cervical tumorigenesis: iASPP acts
Nils Ludwig et al.
Journal of extracellular vesicles, 11(12), e12294-e12294 (2022-12-21)
Transforming growth factor β (TGFβ) is a major component of tumor-derived small extracellular vesicles (TEX) in cancer patients. Mechanisms utilized by TGFβ+ TEX to promote tumor growth and pro-tumor activities in the tumor microenvironment (TME) are largely unknown. TEX produced
Elevated concentrations of serum matrix metalloproteinase-2 and-9 and their associations with circulating markers of cardiovascular diseases in chronic arsenic-exposed individuals.
Islam MS, et al.
Environmental Health : A Global Access Science Source, 14(1), 92-92 (2015)
Matrix metalloproteinases 2 and 9 polymorphism in patients with myeloproliferative diseases: a STROBE-compliant observational study.
Maral S, et al.
Medicine, 94(16) (2015)
Matrix metalloproteinases: old dogs with new tricks.
Somerville RP, et al.
Genome Biology, 4(6), 216-216 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service