Skip to Content
Merck
All Photos(6)

Key Documents

HPA000980

Sigma-Aldrich

Anti-AOC3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HPAO antibody produced in rabbit, Anti-Membrane copper amine oxidase antibody produced in rabbit, Anti-SSAO antibody produced in rabbit, Anti-Semicarbazide-sensitive amine oxidase antibody produced in rabbit, Anti-VAP-1 antibody produced in rabbit, Anti-Vascular adhesion protein 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

DIDQMIFNRELPQASGLLHHCCFYKHRGRNLVTMTTAPRGLQSGDRATWFGLYYNISGAGFFLHHVGLELLVNHKALDPARWTIQKVFYQGRYYDSLAQLEAQFEAGLVNVVLI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AOC3(8639)

Immunogen

Membrane copper amine oxidase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Amine oxidase, copper containing 3 is an enzyme encoded by the AOC3 gene in humans. It is a copper-containing SSAO (semicarbazide-sensitive amine oxidase) secreted by vascular smooth muscle cells, adipocytes and endothelial cells. It is a human endothelial sialoglycoprotein whose cell surface expression is induced under inflammatory conditions. It is a novel adhesion molecule with dual function. It is a proinflammatory protein that helps in mediating leukocyte recruitment through its semicarbazide-sensitive amine oxidase (SSAO) activity. It also participates in inflammation and catalyzes the breakdown of amines to produce aldehyde, hydrogen peroxide and ammonia. Elevation of its activity is observed in atherosclerosis, diabetes mellitus and obesity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76127

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ewa Koc-Zorawska et al.
Postepy higieny i medycyny doswiadczalnej (Online), 67, 1340-1344 (2014-01-01)
VAP-1 (vascular adhesion protein-1) possesses semicarbazide-sensitive amine oxidase (SSAO) activity. It has also been found that serum VAP-1 was elevated in acute and chronic hyperglycemia and in patients with diabetes as well as in chronic kidney disease. Renalase, with possible
Ping Sun et al.
Cerebrovascular diseases (Basel, Switzerland), 37(3), 171-180 (2014-02-08)
In the acute phase of ischemic stroke, endothelial cells are activated and induce the expression of adhesion molecules. Vascular adhesion protein 1 (VAP-1) is a proinflammatory protein that mediates leukocyte recruitment through its semicarbazide-sensitive amine oxidase (SSAO) activity (EC 1.4.3.21).
Eirini Bournazou et al.
International journal of molecular sciences, 20(11) (2019-05-31)
To investigate the expression of vascular adhesion protein-1 (VAP-1) in joint tissues and serum in symptomatic knee osteoarthritis (SKOA) patients and examine whether VAP-1 levels predict increased risk of disease severity in a cross-sectional study. Baseline VAP-1 expression and soluble
Stephen T Ward et al.
BMC cancer, 16, 154-154 (2016-02-26)
The endothelial adhesion molecule, vascular adhesion protein-1 (VAP-1, AOC3) promotes lymphocyte recruitment to tumours, although the contribution that VAP-1 makes to lymphocyte recruitment in human colorectal cancer (CRC) is unknown. VAP-1 exists in circulating soluble form (sVAP-1). A previous study
E Koc-Zorawska et al.
Transplantation proceedings, 45(5), 2009-2012 (2013-06-19)
VAP-1 (vascular adhesion protein-1) is a copper-containing SSAO (semicarbazide-sensitive amine oxidase) secreted by vascular smooth muscle cells, adipocytes, and endothelial cells. Elevation of SSAO activity is observed in atherosclerosis, diabetes mellitus, and obesity. The aim of the study was to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service