Skip to Content
Merck
All Photos(7)

Key Documents

HPA000893

Sigma-Aldrich

Anti-SERPINA3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-α-1-antichymotrypsin, Anti-Aact, Anti-Act, Anti-Serpin peptidase inhibitor, clade a (α-1 antiproteinase, antitrypsin), member 3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:2500-1:5000

immunogen sequence

LASANVDFAFSLYKQLVLKAPDKNVIFSPLSISTALAFLSLGAHNTTLTEILKGLKFNLTETSEAEIHQSFQHLLRTLNQSSDELQLSMGNAMFVKEQLSLLDRFTEDAKRLYGSEAFATDFQDSAAAKKLINDYVKNGGAPHR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SERPINA3(12)

General description

The α1-antichymotrypsin (ACT) protein has a molar mass of 55-66kDa. It has a serpin structure with three β sheets, 8 α helices and a reactive centre loop containing the active site. The gene is mapped to human chromosome 14q32.1.

Immunogen

serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

α1-antichymotrypsin (ACT) is an α-globulin glycoprotein that is a member of the serpin superfamily and is encoded by the SERPINA3 gene in humans. Its expression is higher in prostate cancer than in benign prostatic hyperplasia. The serine protease has been found to be associated with several diseases such as chronic obstructive pulmonary disease (COPD), Parkinson′s disease (PD), Alzheimer′s disease (AD), stroke, cystic fibrosis, cerebral haemorrhage and multiple system atrophy. The encoded protein forms a complex with toxic β-amyloid (Aβ). The gene expression is induced by cytokines. ACT inhibits serine proteases, such as pancreatic chymotrypsin, leukocyte cathepsin G, mast cell chymases, human glandular kallikrein 2, kallikrein 3, and pancreatic cationic elastase. The levels of circulating ACT increases with inflammation. It also has the ability to bind to DNA unlike other serpins.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST82412

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kagenori Ito et al.
Molecular oncology, 17(10), 2147-2167 (2023-07-06)
Clinically, the osteolytic phenotype is rare in prostate cancer (PCa), and the prognosis is generally worse than that of the osteoblastic phenotype. Osteoblastic prostate cancer (BPCa) is a major type of bone metastasis. Several factors responsible for osteogenesis have been
Crystal Baker et al.
Frontiers in bioscience : a journal and virtual library, 12, 2821-2835 (2007-05-09)
Recent advances in proteomic, transcriptomic and genomic technologies have revealed much about the ACT protein and gene. In this review, we summarize our current understanding of the structure and potential physiological roles of the ACT protein, catalogue the regulatory elements
Lei Zhu et al.
The Prostate, 73(2), 219-226 (2012-07-19)
PSA is the most useful prostate cancer marker. However, its levels are increased also in some non-malignant conditions. In circulation, the majority of PSA is complexed with protease inhibitors, including α(1) -antichymotrypsin (ACT). The proportion of the PSA-ACT complex is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service