Skip to Content
Merck
All Photos(5)

Key Documents

HPA000694

Sigma-Aldrich

Anti-DICER1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Dicer, Anti-HERNA, Anti-K12H4.8-LIKE, Anti-KIAA0928, Anti-MNG1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

PTDADSAYCVLPLNVVNDSSTLDIDFKFMEDIEKSEARIGIPSTKYTKETPFVFKLEDYQDAVIIPRYRNFDQPHRFYVADVYTDLTPLSKFPSPEYETFAEYYKTKYNLDLTNLNQPLLDVDHTSSRLNLLTPRHLNQKGKALPLSSAEKRKAKWESLQNKQILVPELCAIHPIPASLWRKAVCLPSILYRLH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DICER1(23405)

Related Categories

General description

Dicer regulates the formation of siRNAs and miRNAs from dsRNA substrates and thereby modulates post-transcriptional silencing of genes. DICER1 depletion in retinal pigmented epithelium has been associated with geographic atrophy, whereas lack of DICER1 in the β cells of pancreas has been linked to diabetes . Anti-DICER1 antibody is specific for DICER1 in humans.

Immunogen

Endoribonuclease Dicer recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
Western Blotting (1 paper)

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70370

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole S Spoelstra et al.
Human pathology, 56, 40-51 (2016-06-05)
Dicer is an RNase III enzyme responsible for cleaving double-stranded RNAs into small interfering RNAs and microRNAs, which either target messenger RNA transcripts for degradation or inhibit translation. Dicer protein levels have been examined in breast cancer with contradictory results.
Idoia Martínez de LaPiscina et al.
Frontiers in endocrinology, 11, 433-433 (2020-07-28)
Context: The DICER1 syndrome is a multiple neoplasia disorder caused by germline mutations in the DICER1 gene. In DICER1 patients, aggressive congenital pituitary tumors lead to neonatal Cushing's disease (CD). The role of DICER1 in other corticotropinomas, however, remains unknown.
Bruno Vincenzi et al.
European journal of cancer (Oxford, England : 1990), 49(6), 1501-1508 (2012-12-26)
The miRNA-regulating enzymes Dicer and Drosha exhibit aberrant expression in several cancer types. Dicer and Drosha play a crucial role during the angiogenetic process in vitro and, for Dicer, in vivo. We aimed to investigate the potential role of Dicer
Shweta Dambal et al.
The Journal of steroid biochemistry and molecular biology, 167, 192-202 (2017-01-17)
Vitamin D deficiency increases the risk of lethal prostate adenocarcinomas (PCa) and the majority of older men are deficient. Although PCa arises from the epithelium, the surrounding stroma has hormonal regulatory control over the epithelium and contributes to carcinogenesis. Herein
Miseon Lee et al.
Pathobiology : journal of immunopathology, molecular and cellular biology, 88(3), 251-260 (2021-02-11)
Pleuropulmonary blastoma (PPB) is a rare sarcomatous malignancy involving the lung and pleura which occurs in early childhood. Cystic PPB in the early stage can be misdiagnosed as other cystic diseases. Early detection of this entity is important for appropriate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service