Skip to Content
Merck
All Photos(4)

Documents

HPA000496

Sigma-Aldrich

Anti-CCNB3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-G2/mitotic-specific cyclin-B3 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

KMCASQRKQSCQEESLAVQDVNMEEDSFFMESMSFKKKPKTEESIPTHKLSSLKKKCTIYGKICHFRKPPVLQTTICGAMSSIKKPTTEKETLFQELSVLQEKHTTEHEMS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCNB3(85417)

Immunogen

G2/mitotic-specific cyclin-B3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74152

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tricia L Peters et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 28(4), 575-586 (2014-11-02)
The BCOR-CCNB3 fusion gene, resulting from a chromosome X paracentric inversion, was recently described in translocation-negative 'Ewing-like' sarcomas arising in bone and soft tissue. Genetic subclassification of undifferentiated unclassified sarcomas may potentially offer markers for reproducible diagnosis and substrates for
Akihiko Yoshida et al.
Histopathology, 76(4), 509-520 (2019-10-28)
BCOR gene alteration is a genetic signature of rare subsets of sarcomas. Most BCOR-associated sarcomas thus far reported are in the pediatric population, except for uterine sarcomas. We studied seven cases of BCOR-associated non-uterine sarcomas in adult patients. The patients
Kayo Suzuki et al.
Molecular and clinical oncology, 16(4), 77-77 (2022-03-08)
BCL6 corepressor-cyclin B3 (BCOR-CCNB3) fusion sarcoma was classified as an emerging subgroup of undifferentiated small round cell sarcoma in 2020. The incidence of BCOR-CCNB3 fusion sarcoma is reportedly 1.5-14% among undifferentiated unclassified sarcomas, representing a rare entity among primary malignant
Daisuke Ichikawa et al.
NPJ genomic medicine, 6(1), 49-49 (2021-06-17)
Pediatric solid tumors are a heterogeneous group of neoplasms with over 100 subtypes. Clinical and histopathological diagnosis remains challenging due to the overlapping morphological and immunohistochemical findings and the presence of atypical cases. To evaluate the potential utility of including
Meng K Wong et al.
Histopathology, 72(2), 320-329 (2017-08-24)
Clear cell sarcoma of the kidney (CCSK) is a rare paediatric renal malignant tumour. The majority of CCSKs have internal tandem duplications (ITDs) of the BCOR gene, whereas a minority have the YWHAE-NUTM2 gene fusion. A third 'double-negative' (DN) category

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service