Skip to Content
Merck
All Photos(1)

Key Documents

H9914

Millipore

HIS-Select® Nickel Magnetic Agarose Beads

Synonym(s):

nickel charged magnetic beaded agarose

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

conjugate

magnetic beads

form

suspension

shelf life

2 yr (Unopened product)

matrix

6% beaded magnetic agarose

capacity

≥15 mg/mL binding capacity

storage temp.

2-8°C

General description

HIS-Select Magnetic Agarose Beads consist of paramagnetic, immobilized metal-ion affinity chromatography (IMAC) resin that contain a proprietary quadridentate chelate, which is bound with nickel and covalently attached through a non-charged, hydrophilic linker to magnetic beaded agarose. The magnetic properties of the beads aid in manipulations, such as repetitive washings, and recovery of the protein bound beads. This leads to greater experimental reproducibility and more accurate quantitation of the His-tagged proteins of interest.

Application

The HIS-Select Nickel Magnetic Agarose Beads are designed for use in automated and small-scale affinity capture (molecular pull-down) purifications of histidine-tagged protein / His-tag protein while exhibiting low non-specific binding of other proteins. The beads can be used to purify His-tagged proteins under native and denaturing conditions.

Physical form

Supplied as a 50% slurry suspension in 30% Ethanol.

Legal Information

HIS-Select is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Flam. Liq. 3 - Repr. 1B - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

86.0 °F

Flash Point(C)

30 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonja Hänzelmann et al.
Clinical epigenetics, 7, 19-19 (2015-03-13)
Primary cells enter replicative senescence after a limited number of cell divisions. This process needs to be considered in cell culture experiments, and it is particularly important for regenerative medicine. Replicative senescence is associated with reproducible changes in DNA methylation
Mei Suen Kong et al.
Science signaling, 12(567) (2019-02-07)
T cell activation is initiated by signaling molecules downstream of the T cell receptor (TCR) that are organized by adaptor proteins. CIN85 (Cbl-interacting protein of 85 kDa) is one such adaptor protein. Here, we showed that CIN85 limited T cell
Philip Vitorino et al.
Nature, 519(7544), 425-430 (2015-03-25)
Cell migration is a stepwise process that coordinates multiple molecular machineries. Using in vitro angiogenesis screens with short interfering RNA and chemical inhibitors, we define here a MAP4K4-moesin-talin-β1-integrin molecular pathway that promotes efficient plasma membrane retraction during endothelial cell migration.
Joshua W Burgess et al.
American journal of respiratory cell and molecular biology, 41(6), 714-721 (2009-03-17)
Pneumocystis organisms are opportunistic fungal pathogens that cause significant pneumonia in immune-compromised hosts. Recent evidence has suggested that Pneumocystis carinii exists as separate mating types, and expresses and regulates proteins that govern meiosis and progression of the life cycle. This
Yun Wei et al.
Journal of materials chemistry. B, 1(15), 2066-2071 (2013-04-21)
The imidazolium cation was chosen as protein selective affinity group and a kind of ionic liquid magnetic microspheres was developed by immobilizing imidazolium cations onto the surface of silica-coated magnetic microspheres to form imidazolium ionic liquid modified magnetic microspheres (Fe3O4@SiO2@IL).

Related Content

Protein purification techniques, reagents, and protocols for purifying recombinant proteins using methods including, ion-exchange, size-exclusion, and protein affinity chromatography.

Protein purification techniques, reagents, and protocols for purifying recombinant proteins using methods including, ion-exchange, size-exclusion, and protein affinity chromatography.

Protein purification techniques, reagents, and protocols for purifying recombinant proteins using methods including, ion-exchange, size-exclusion, and protein affinity chromatography.

Protein purification techniques, reagents, and protocols for purifying recombinant proteins using methods including, ion-exchange, size-exclusion, and protein affinity chromatography.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service