Skip to Content
Merck
All Photos(2)

Key Documents

H9390

Sigma-Aldrich

Hyaluronic acid sodium salt from Streptococcus zooepidemicus

bacterial glycosaminoglycan polysaccharide

Synonym(s):

Poly(β-glucuronic acid-[1→3]-β-N-acetylglucosamine-[1→4]), alternating

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

bacterial (Streptococcus zooepidemicus)

Quality Level

form

powder

color

white

solubility

water: 20 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

[Na+].N([C@H]1[C@@H](O[C@@H]([C@H]([C@@H]1O[C@@H]4O[C@@H]([C@H]([C@@H]([C@H]4O)O)O)C(=O)O)O)CO)O[C@@H]2[C@H](O[C@H]([C@@H]([C@H]2O)O)O[C@@H]3[C@H]([C@@H](O[C@@H]([C@H]3O)CO)O)NC(=O)C)C(=O)O)C(=O)C

InChI

1S/C28H44N2O23.Na/c1-5(33)29-9-18(11(35)7(3-31)47-25(9)46)49-28-17(41)15(39)20(22(53-28)24(44)45)51-26-10(30-6(2)34)19(12(36)8(4-32)48-26)50-27-16(40)13(37)14(38)21(52-27)23(42)43;/h7-22,25-28,31-32,35-41,46H,3-4H2,1-2H3,(H,29,33)(H,30,34)(H,42,43)(H,44,45);/q;+1/t7-,8-,9-,10-,11-,12-,13+,14+,15-,16-,17-,18-,19-,20+,21+,22+,25-,26+,27-,28-;/m1./s1

InChI key

YWIVKILSMZOHHF-QJZPQSOGSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalie Fiander
British journal of nursing (Mark Allen Publishing), 22(9), S26-S26 (2013-06-12)
Painful bladder syndrome (PBS) and interstitial cystitis (IC) are associated with bladder pain, increased urinary frequency, urgency and reduced quality of life. The cause is still unknown, although there are several possible hypotheses. PBS/IC may significantly impact all areas of
Treatment of hyaluronan accumulation ameliorates high-fat diet-induced insulin resistance in mice.
Per Fogelstrand et al.
Diabetes, 62(6), 1816-1817 (2013-05-25)
Shenglian Yao et al.
Journal of nanoscience and nanotechnology, 13(7), 4752-4758 (2013-08-02)
Nanofibers exist ubiquitously in natural extracellular matrix (ECM) of all kinds of human tissues forming hydrated interwoven network. Electrospinning nanotechnology has been proven to be a powerful technique to fabricate controllable nanofibers mimicking the natural ECM structures. Hyaluronic acid (HA)
Daniela Elena Costea et al.
Cancer research, 73(13), 3888-3901 (2013-04-20)
Heterogeneity of carcinoma-associated fibroblasts (CAF) has long been recognized, but the functional significance remains poorly understood. Here, we report the distinction of two CAF subtypes in oral squamous cell carcinoma (OSCC) that have differential tumor-promoting capability, one with a transcriptome
Incorporating facial rejuvenation procedures in the dental practice.
Warren Roberts
Today's FDA : official monthly journal of the Florida Dental Association, 25(2), 50-53 (2013-05-23)

Questions

1–2 of 2 Questions  
  1. I am currently using hyaluronic acid sodium salt (cat no: H9390, lot no: 120k15211). Can I know the Intrinsic Viscosity of this material?

    1 answer
    1. Intrinsic viscosity is not a property measured or reported for Product H9390.

      Helpful?

  2. 分子量を教えてください

    1 answer
    1. Hyaluronic acid from this source has a molecular weight range of 0.85 to 1.6 million Daltons.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service