Skip to Content
Merck
All Photos(5)

Key Documents

H9268

Sigma-Aldrich

Hexadimethrine bromide

≥94% (titration)

Synonym(s):

1,5-Dimethyl-1,5-diazaundecamethylene polymethobromide, Polybrene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106502
NACRES:
NA.26
Pricing and availability is not currently available.

Assay

≥94% (titration)

storage temp.

2-8°C

SMILES string

BrCCCBr.N(CCCCCCN(C)C)(C)C

InChI

1S/C10H24N2.C3H6Br2/c1-11(2)9-7-5-6-8-10-12(3)4;4-2-1-3-5/h5-10H2,1-4H3;1-3H2

InChI key

KZKAYEGOIJEWQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hexadimethrine bromide is a quaternary ammonium compound which works as a heparin antagonist.[1] It is also involved in the transformation of low molecular weight DNA.[2]

Application

Hexadimethrine bromide can be used to transfect mammalian cells with DNA. It can be used to increase the efficiency of lipofection transfections.
Hexadimethrine bromide has been used for lentivirus infection in cells.[3][4] It has been used for infection of cells with retroviral supernatant.[5][6]

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Redei GP
Encyclopedia of Genetics, Genomics, Proteomics, and Informatics (2008)
UCHL1 Is a Putative Tumor Suppressor in Ovarian Cancer Cells and Contributes to Cisplatin Resistance.
Jin C et al.
Journal of Cancer, 4, 662-662 (2013)
Blood (2013)
Gaetana Restivo et al.
Nature communications, 8(1), 1988-1988 (2017-12-08)
Cutaneous melanoma represents the most fatal skin cancer due to its high metastatic capacity. According to the "phenotype switching" model, the aggressive nature of melanoma cells results from their intrinsic potential to dynamically switch from a high-proliferative/low-invasive to a low-proliferative/high-invasive
An important role of matrix metalloproteinase-8 in angiogenesis in vitro and in vivo.
Fang C et al.
Cardiovascular Research, 99, 146-146 (2013)

Articles

Small interfering RNAs (siRNAs) are powerful tools for gene expression knockdown, widely used in molecular biology.

Small interfering RNAs (siRNAs) are powerful tools for gene expression knockdown, widely used in molecular biology.

Small interfering RNAs (siRNAs) are powerful tools for gene expression knockdown, widely used in molecular biology.

Small interfering RNAs (siRNAs) are powerful tools for gene expression knockdown, widely used in molecular biology.

Protocols

Detailed procedure for how to perform a lentiviral transduction of MISSION shRNA lentiviral particles to achieve a stable long term silencing and phenotypic change.

Detailed procedure for how to perform a lentiviral transduction of MISSION shRNA lentiviral particles to achieve a stable long term silencing and phenotypic change.

Detailed procedure for how to perform a lentiviral transduction of MISSION shRNA lentiviral particles to achieve a stable long term silencing and phenotypic change.

Detailed procedure for how to perform a lentiviral transduction of MISSION shRNA lentiviral particles to achieve a stable long term silencing and phenotypic change.

See All

Questions

1–10 of 12 Questions  
  1. Is it possible for hexadimethrine bromide to breakdown and lose its effectiveness after 3 years when stored in its powdered form at 4 degree celsius? We bought this product around 3 years and now it doesn't work as well as before when doing transduction.

    1 answer
    1. This product is assigned a recommended retest period of 2 years from the date of Quality release. While quite stable, the compound may slowly degrade over time. The suitability of this item beyond the lot specific retest date would have to be determined by the end user.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  4. what is the concentration of 5G polybrene? Does G stands for gram?

    1 answer
    1. This product is in powder form and not a prepared solution. The minimum purity of this material is 94%. The 'G' represents grams.

      Helpful?

  5. What is the preferred solvent for Hexadimethrine bromide? I see water or saline above, but does either effect solubility or stability?

    1 answer
    1. The choice of solvent typically depends on the specific application. For in vitro or in vivo studies, saline is more commonly used. The material is soluble in water at up to 10%. Solutions are quite stable and maintain activity after autoclaving. Stock solutions prepared at 1 mg/mL in saline are stable for up to 1 year when stored at 2–8 °C. Please see the link below to review additional information available in the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/253/063/h9268bul.pdf

      Helpful?

  6. How long is the Hexadimethrine Bromide powder stable? I cannot find its expiry date.

    1 answer
    1. This product in powdered format is stable for up to a recommended retest period of two years as listed on the Certificate of Analysis. After the Retest Date, product samples should be examined to ensure that the product is still in compliance with the established specifications.

      When in solution, this product will remain active for at least one year if stored at 2–8 °C. For more information please view the product data sheet: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/253/063/h9268bul.pdf

      Helpful?

  7. Is Product H9268, Hexadimethrine bromide, the same thing as Polybrene?

    1 answer
    1. Yes, Polybrene (registered trade-mark of Abbott) and 1,5-Dimethyl-1,5-diazaundecamethylene polymethobromide, are Synonyms.

      Helpful?

  8. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  9. What is the molecular weight of Product H9268, Hexadimethrine bromide?

    1 answer
    1. According to the information from our supplier, the molecular weight of this product is 4,000-6,000.The chemicals encyclopedia published by the Royal Society of Chemistry provides a general MW range for the chemical of 5,000 - 10,000.

      Helpful?

  10. How do I sterilize Product H9268, Hexadimethrine bromide?

    1 answer
    1. Hexadimethrine bromide can be filtered sterilized or autoclaved.

      Helpful?

1–10 of 12 Questions  

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service