Skip to Content
Merck

Skip To

H8627

Hydroxyurea

98% (elemental analysis), powder, RNR inhibitor

Synonym(s):

Hydroxycarbamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
NH2CONHOH
CAS Number:
Molecular Weight:
76.05
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
204-821-7
MDL number:
Beilstein/REAXYS Number:
1741548
Assay:
98%
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Hydroxyurea, 98%, powder

Quality Segment

assay

98%

form

powder

color

white

solubility

H2O: 50 mg/mL

storage temp.

2-8°C

SMILES string

NC(=O)NO

InChI

1S/CH4N2O2/c2-1(4)3-5/h5H,(H3,2,3,4)

InChI key

VSNHCAURESNICA-UHFFFAOYSA-N

General description

Hydroxyurea is a hydroxylated analog of urea. It is beneficial in increasing fetal hemoglobin (HbF) levels, which in turn reduces the frequency of severe crises and the need for blood transfusions in sickle cell anemia patients. Hydroxyurea exhibits cell-cycle specificity for the S phase, leading to cell arrest at G1 to S. This non-alkylating S phase-specific myelosuppressive agent has a mechanism of action that is not specific to platelets.

Application

Hydroxyurea has been used:
  • for cell cycle synchronization in MCF-7 human breast carcinoma cells, Schizosaccharomyces pombe, primary root of seedling
  • as a drug for treating the parasite Leishmania mexicana in cell viability and cell cycle assay
  • as an inhibitor in the cell adhesion and motility assay in Caki control and CD9 knockdown cells

Biochem/physiol Actions

Anti-neoplastic. Inactivates ribonucleoside reductase by forming a free radical nitroxide that binds a tyrosyl free radical in the active site of the enzyme. This blocks the synthesis of deoxynucleotides, which inhibits DNA synthesis and induces synchronization or cell death in S-phase.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR2629400046Y0000119
form

powder

form

solid

form

solid

form

-

assay

98%

assay

-

assay

≥98% (HPLC)

assay

-

Quality Level

200

Quality Level

300

Quality Level

100

Quality Level

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

H2O: 50 mg/mL

solubility

-

solubility

water: 5 mg/mL

solubility

-

color

white

color

-

color

white

color

-


Still not finding the right product?

Explore all of our products under Hydroxyurea


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Muta. 1B - Repr. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. How long is Hydroxyurea stable for once resuspended in water? Still store at 2-8C once resuspended? Is it sterile (suitable for cell culture work)?

    1 answer
    1. Hydroxyurea decomposes in the presence of moisture, making aqueous solutions likely unstable. Solutions should be prepared immediately before use. This product is not sterile.

      Helpful?

Reviews

No rating value

Active Filters