Skip to Content
Merck
All Photos(1)

Key Documents

H6287

Sigma-Aldrich

Anti-Histone Deacetylase 1 (HDAC1) antibody, Mouse monoclonal

clone HDAC1-21, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-Histone Deacetylase 1 (HDAC1) antibody produced in mouse

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HDAC1-21, monoclonal

form

buffered aqueous solution

mol wt

antigen ~65 kDa

species reactivity

mouse, human

concentration

~2 mg/mL

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
microarray: suitable
western blot: 2-4 μg/mL using total cell extracts of HeLa cells

isotype

IgG3

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HDAC1(3065)
mouse ... Hdac1(433759)

General description

Anti-Histone Deacetylase 1 (HDAC1) antibody, Mouse monoclonal (mouse IgG3 isotype) is derived from the HDAC1-21 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to amino acids of human and mouse histone deacetylase 1 (HDAC1).
Histone deacetylases (HDACs) are competing enzymes, belonging to histone deacetylase family. There are two classes of HDACs with six to seven different types of HDACs proteins. HDAC1,HDAC2 and HDAC3 belong to Class I HDACs and HDAC4, HDAC6, and HDAC7 belong to Class II HDACs. Class I HDACs consists of a single deacetylase domain at the N-termini and diversified C-terminal regions, while Class II contains a deacetylase domain at C-terminal position.

Application

Anti-Histone Deacetylase 1 (HDAC1) antibody, Mouse monoclonal has been used in:
  • enzyme-linked immunosorbent assay (ELISA)
  • immunoprecipitation
  • immunoblotting

Biochem/physiol Actions

HDAC1 is widely studied and is shown to actively modulate the eukaryotic chromatin structure. It deacetylyses lysine residues on core histones (H2A, H2B, H3 and H4) at the N-terminal. It is a vital component of cofactor complexes and is involved in transcription regulation. Histone deacetylation results in transcription repression leading to the formation of tight nucleosomal structure which prevents DNA accessing.

Physical form

solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stem cell factor SALL4 represses the transcriptions of PTEN and SALL1 through an epigenetic repressor complex
Lu J, et al.
PLoS ONE, 4(5), e5577-e5577 (2009)
Engineering cell signaling using tunable CRISPR-Cpf1-based transcription factors
Liu Y, et al.
Nature Communications, 8(1), 2095-2095 (2017)
Coactivators and corepressors of NF-kappaB in IkappaB alpha gene promoter
Gao Z, et al.
The Journal of biological chemistry, 280(22), 21091-21098 (2005)
Sudhakar Ammanamanchi et al.
The Journal of biological chemistry, 278(37), 35775-35780 (2003-07-03)
Sp3 transcription factor can either activate or repress target gene expression. However, the molecular event that controls this dual function is unclear. We previously reported (Ammanamanchi, S., and Brattain, M. G. (2001) J. Biol. Chem. 276, 3348-3352) that unmodified Sp3
Jiayun Lu et al.
PloS one, 4(5), e5577-e5577 (2009-05-15)
The embryonic stem cell (ESC) factor, SALL4, plays an essential role in both development and leukemogenesis. It is a unique gene that is involved in self-renewal in ESC and leukemic stem cell (LSC). To understand the mechanism(s) of SALL4 function(s)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service