Skip to Content
Merck
All Photos(1)

Key Documents

H6030

Sigma-Aldrich

Anti-HSV antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Herpes Simplex Virus

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.56

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

immunoprecipitation (IP): 1.0 μg/mL
western blot: 2.5 μg/mL

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Recognizes N- and C-terminal HSV fusion proteins.

Immunogen

synthetic peptide corresponding to amino acids 290−300 of glycoprotein-D precursor, an envelop component of herpes simplex virus.

Application

Anti-HSV antibody was used to expand the repertoire of plasmids for PCR-mediated epitope tagging in yeast.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Chromatin immunoprecipitation (1 paper)
Western Blotting (1 paper)

Biochem/physiol Actions

Anti-HSV is developed in rabbits using a synthetic peptide (K-QPELAPEDPED) conjugated to KLH via the N-terminal lysine. The peptide corresponds to amino acids 290-300 of Glycoprotein D precursor which is an envelope component of herpes simplex virus. Anti-HSV antibody reacts specifically with HSV tagged fusion proteins, using immunoblotting and immunoprecipitation techniques.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expanding the repertoire of plasmids for PCR-mediated epitope tagging in yeast
Moqtaderi Z, Struhl K
Yeast, 4, 287-292 (2008)
Obstetrical roundabout.
H E Cudby
Midwives chronicle, 91(1089), 301-301 (1978-10-01)
Christine E Cucinotta et al.
Nucleic acids research, 47(16), 8410-8423 (2019-06-22)
The nucleosome core regulates DNA-templated processes through the highly conserved nucleosome acidic patch. While structural and biochemical studies have shown that the acidic patch controls chromatin factor binding and activity, few studies have elucidated its functions in vivo. We employed
P O Olins et al.
Current opinion in biotechnology, 4(5), 520-525 (1993-10-01)
Recent advances in protein expression in E. coli have focused primarily on the enhancement of protein quality. Problems in mRNA translation such as inefficient initiation, mistranslation, frame-shifting and frame-hopping can often be addressed by altering heterologous gene-coding sequences. Fusion technology
Mitchell A Ellison et al.
Nucleic acids research, 51(10), 4814-4830 (2023-03-18)
The Paf1 complex (Paf1C) is a conserved transcription elongation factor that regulates transcription elongation efficiency, facilitates co-transcriptional histone modifications, and impacts molecular processes linked to RNA synthesis, such as polyA site selection. Coupling of the activities of Paf1C to transcription

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service