Skip to Content
Merck
All Photos(1)

Documents

H4663

Sigma-Aldrich

HMBA

≥98% (HPLC), solid

Synonym(s):

10-[(3-Hydroxy-4-methoxybenzylidene)]-9(10H)-anthracenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H16O3
CAS Number:
Molecular Weight:
328.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: soluble 34 mg/mL

storage temp.

−20°C

SMILES string

COc1ccc(cc1O)\C=C2\c3ccccc3C(=O)c4ccccc24

InChI

1S/C22H16O3/c1-25-21-11-10-14(13-20(21)23)12-19-15-6-2-4-8-17(15)22(24)18-9-5-3-7-16(18)19/h2-13,23H,1H3

InChI key

MQLACMBJVPINKE-UHFFFAOYSA-N

Biochem/physiol Actions

HMBA is an Inhibitor of microtubule polymerization; apoptosis inducer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Wang et al.
Pharmaceutical research, 31(12), 3538-3548 (2014-06-20)
To determine the effect of PEG modification on pharmacologic and gene delivery properties of polymeric CXCR4 antagonist based on Plerixafor. Polymeric Plerixafor (PAMD) was synthesized from Plerixafor (AMD3100) and grafted with different amounts of PEG (2 kDa). CXCR4 antagonism of the
Discovery of a large-scale, cell-state-responsive allosteric switch in the 7SK RNA using DANCE-MaP.
Olson, et al.
Molecular Cell, 82, 1708-1723 (2023)
Erwan Dupuis et al.
Talanta, 219, 121345-121345 (2020-09-06)
Capillary electrophoresis (CE) was hyphenated to multi-collector inductively coupled plasma mass spectrometry (MC-ICP-MS) to determine the model age of a highly enriched uranium (HEU) sample using the 234U/230Th radiochronometer. The use of hydroxymethylbutyric acid (HMBA) as the CE electrolyte was
Seung Han Son et al.
International journal of molecular sciences, 23(12) (2022-06-25)
CP2c, also known as TFCP2, α-CP2, LSF, and LBP-1c, is a prototypic member of the transcription factor (TF) CP2 subfamily involved in diverse ubiquitous and tissue/stage-specific cellular processes and in human malignancies including cancer. Despite its importance, many fundamental regulatory
Dong-Hui Xu et al.
International journal of oncology, 45(1), 264-272 (2014-05-03)
Nucleophosmin (NPM, also known as B23), mainly localized in the nucleolus, has been reported to be overexpressed in many types of human cancer, including colon, ovarian, prostate and gastric cancer. NPM was identified while screening the differential nuclear matrix proteins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service