Skip to Content
Merck
All Photos(1)

Key Documents

H4535

Sigma-Aldrich

Anti-Heme Oxygenase-1 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-HO-1, Anti-HSP32

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~32 kDa

species reactivity

rat, mouse

technique(s)

immunohistochemistry: 5 μg/mL
immunoprecipitation (IP): 10-20 μg/mL
western blot: 0.5-2 μg/mL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HMOX1(3162)
mouse ... Hmox1(15368)
rat ... Hmox1(24451)

Immunogen

Synthetic peptide surrounding amino acids near the N-terminus of human HO-1.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)
Western Blotting (1 paper)

Physical form

Solution in phosphate buffered saline containing 30% glycerol, 0.5% BSA and 0.01% thimerosal.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorenzo L F Glorie et al.
American journal of physiology. Renal physiology, 303(5), F681-F688 (2012-06-22)
Dipeptidyl peptidase 4 (DPP4) is an exopeptidase which modulates the function of its substrates, among which are insulin-releasing incretins. DPP4 inhibitors are currently used to improve glucose tolerance in type 2 diabetes patients. Inhibition of DPP4 exhibits protective effects on
Megan A Mahlke et al.
Pathobiology of aging & age related diseases, 1 (2011-01-01)
The anti-tumor effects of calorie restriction (CR) and the possible underlying mechanisms were investigated using ethylnitrosourea (ENU)-induced glioma in rats. ENU was given transplacentally at gestational day 15, and male offspring were used in this experiment. The brain from 4-
Abdulla K Salahudeen et al.
Transplantation, 91(2), 176-182 (2010-11-16)
Kidneys from brain-dead donors are cold preserved until transplanted. However, prolonged cold storage can contribute to allograft failure. Studies suggest that donor preconditioning with dopaminergics may reduce cold-ischemic transplant injury, but whether heme oxygenase (HO)-1 induction is an underlying mechanism
Qunli Cui et al.
Molecular medicine reports, 13(2), 1381-1388 (2015-12-10)
Parkinson's disease (PD) is an age-related complex neurodegenerative disease that affects ≤ 80% of dopaminergic neurons in the substantia nigra pars compacta (SNpc). It has previously been suggested that mitochondrial dysfunction, oxidative stress and oxidative damage underlie the pathogenesis of
Alexander Leathem et al.
Antioxidants (Basel, Switzerland), 11(2) (2022-02-26)
The cyclic nitroxide TEMPOL exerts anti-oxidative and anti-inflammatory effects, and thus may provide therapeutic benefit in Parkinson's disease (PD), in which mitochondrial dysfunction, oxidative damage and inflammation have been implicated as pathophysiological mechanisms underlying the selective loss of dopaminergic neurons.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service