Skip to Content
Merck

H3261

2-Mercaptopyridine N-oxide sodium salt

97.0-103.0%

Synonym(s):

Sodium Omadine, Sodium pyrithione, 1-Hydroxy-2-pyridinethione sodium salt, 2-Mercaptopyridine-1-oxide sodium salt, 2-Pyridinethiol-1-oxide sodium salt, Pyrithione sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H4NNaOS
CAS Number:
Molecular Weight:
149.15
UNSPSC Code:
51102829
NACRES:
NA.85
PubChem Substance ID:
EC Number:
223-296-5
Beilstein/REAXYS Number:
4026050
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

biological source

synthetic

assay

97.0-103.0%

color

faintly beige, white to light yellow

solubility

H2O: 50 mg/mL

antibiotic activity spectrum

fungi

mode of action

cell membrane | interferes

SMILES string

[Na+].[O-][n+]1ccccc1[S-]

InChI

1S/C5H5NOS.Na/c7-6-4-2-1-3-5(6)8;/h1-4,8H;/q;+1/p-1

InChI key

WNGMMIYXPIAYOB-UHFFFAOYSA-M

Application

2-Mercaptopyridine N-oxide can be used to generate bidentate oxothiolane chelates with transition metals.
2-Mercaptopyridine N-oxide, also called pyrithione, is added to cells in culture so metals enter them readily without relying on transporters. It is used to study picornavirus infections.

Biochem/physiol Actions

2-Mercaptopyridine N-oxide is a zinc ionophore that transports zinc into cells.


Still not finding the right product?


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1

target_organs

Nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library