Skip to Content
Merck

H2292

Histone H3.1 human

Synonym(s):

Histone, Human

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
NACRES:
NA.25
UNSPSC Code:
12352202
Form:
liquid
Biological source:
human
Recombinant:
expressed in E. coli
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

recombinant

expressed in E. coli

form

liquid

concentration

1 mg/mL

technique(s)

MALDI-TOF: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... H3C1(8350)

General description

Histone H3.1 is a histone H3 variant. It differs from H3.2 with residue 96 being a cysteine instead of serine. H3.1 is a replication-dependent histone and is a component of nucleosomes during the S-phase.

Application

Histone H3.1 human has been used in in vitro Schizosaccharomyces pombe Set7 histone methyltransferase assay for matrix-assisted laser desorption ionization-time of flight mass spectrometry (MALDI-TOF) studies.

Biochem/physiol Actions

Histone H3.1 human (H3.1) undergoes a majority of post-translational modification especially acetylation and demethylation in lysine 14 and 9 respectively. During differentiation, the levels of H3.1 transcripts decrease as cell division slows down. The incorporation of H3.1 into chromatin is mediated by a chaperone, chromatin assembly factor (CAF-1). A transversion mutation in the H3.1 may be implicated in glioma.

Physical form

Supplied as 1 mg/mL in 20 mM NaPO4, pH 7.0, 0.3 M NaCl, 1 mM EDTA, and 1 mM DTT.


pictograms

Exclamation mark

signalword

Warning

hcodes

flash_point_f

Not applicable

flash_point_c

Not applicable

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library