Skip to Content
Merck

Skip To

H0887

HEPES solution

1M, liquid, pH 7.0-7.6, suitable for cell culture, BioXtra, BioReagent

HEPES solution

Synonym(s):

N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12161704
PubChem Substance ID:
NACRES:
NA.75
MDL number:
eCl@ss:
32129211
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture

sterility

sterile-filtered

Quality Level

product line

BioReagent, BioXtra

assay

0.9—1.1 M

form

liquid

storage condition

(Tightly closed. Dry), dry at room temperature

concentration

1 M

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

colorless

pH

7.0-7.6

useful pH range

6.8-8.2

 

7.5

density

1.1 g/cm3

suitability

suitable for component for culture media, suitable for electrophoresis

application(s)

advanced drug delivery
cell analysis
life science and biopharma
sample preparation

storage temp.

2-8°C

SMILES string

OCCN1CCN(CCS(=O)(O)=O)CC1

InChI

1S/C8H18N2O4S/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14/h11H,1-8H2,(H,12,13,14)

InChI key

JKMHFZQWWAIEOD-UHFFFAOYSA-N

General description

HEPES Buffer Solution, a sterile-filtered 1M solution of 4-(2-hydroxyethyl)piperazine-1-ethane-sulfonic acid, serves as a vital biological buffer in cell biology, biochemical, and biological research. This zwitterionic buffer enjoys widespread adoption due to its effective pH range of 6.8 to 8.2, making it suitable for a diverse array of research applications. HEPES excels as a buffering agent in cell culture media, offering several advantages over alternatives like Tris and phosphate. Notably, HEPES exhibits lower cell toxicity, ensuring optimal cell health and viability during experiments.

Furthermore, it surpasses bicarbonate buffers (NaHCO3) in its ability to maintain physiological pH despite fluctuations in carbon dioxide concentration arising from cellular respiration, a key factor in maintaining cellular homeostasis. Beyond its exceptional performance in cell culture, HEPES may also find application in various biological and biochemical studies. It may also be valuable in techniques like immunoprecipitation, cell lysis, and live cell imaging, providing a stable and controlled environment for experimental procedures. The combination of its versatility, exceptional pH buffering capacity, and minimal interaction with other molecules positions HEPES as an indispensable tool across diverse research fields.
It is typically used in cell culture at concentration between 5mM to 30 mM. HEPES has been used in a wide variety of applications, including tissue culture. It is commonly used to buffer cell culture media in air. HEPES finds its usage in in vitro experiments on Mg.

Application

HEPES has been used as a component of:
  • Hank’s balanced salt solution, Dulbecco’s modified eagle’s medium and no-glucose DMEM, which are used for the preparation of tissue slices[1]
  • Buffers used in localized conductance measurements on the basolateral side of culture inserts to examine cation selectivity[1]
  • cell culture media to provide buffer capacity to maintain the pH of the cell line[2]
  • buffer in decellularization of porcine nucleus pulposus[3]

Features and Benefits

  • High purity product for biochemical and biological research
  • Sterile filtered and cell culture tested
  • Ready Solution reduces the need
  • Useful for the pH range of 6.8 - 8.2 and pKa of 7.5 at 25 °C
  • Ready available solution reduce the need for preparation time

Preparation Note

Prepared from HEPES free acid and cell culture grade water.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Disclaimer

Concentrations of HEPES over 40mM may be toxic to some cell types.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8326483264H23830
assay

0.9—1.1 M

assay

-

assay

-

assay

≥99% (titration)

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

-

technique(s)

-

concentration

1 M

concentration

1 M in H2O, 238 g/L HEPES in H2O

concentration

1 M in H2O, 238 g/L HEPES in H2O

concentration

-

Quality Level

300

Quality Level

100

Quality Level

100

Quality Level

200

product line

BioReagent, BioXtra

product line

-

product line

-

product line

-

application(s)

cell analysis
life science and biopharma
sample preparation

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

application(s)

-

assay

0.9—1.1 M

assay

-

assay

-

assay

≥99% (titration)

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

-

technique(s)

-

concentration

1 M

concentration

1 M in H2O, 238 g/L HEPES in H2O

concentration

1 M in H2O, 238 g/L HEPES in H2O

concentration

-

Quality Level

300

Quality Level

100

Quality Level

100

Quality Level

200

product line

BioReagent, BioXtra

product line

-

product line

-

product line

-

application(s)

cell analysis
life science and biopharma
sample preparation

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

application(s)

-


Still not finding the right product?

Explore all of our products under HEPES solution


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Product Information Sheet


Kaixi Tang et al.
PLoS computational biology, 14(7), e1006303-e1006303 (2018-07-06)
Focal adhesions are protein complexes that anchor cells to the extracellular matrix. During migration, the growth and disassembly of these structures are spatiotemporally regulated, with new adhesions forming at the leading edge of the cell and mature adhesions disassembling at
Assessing the corrosion of biodegradable magnesium implants: a critical review of current methodologies and their limitations.
Kirkland Nt
Acta Biomaterialia, 8, 925- 936 (2012)
The role of the endocrine system in feeding-induced tissue-specific circadian entrainment.
Sato M, et al.
Cell Reports, 8(2), 393-401 (2014)



Global Trade Item Number

SKUGTIN
H0887-20ML04061838055101
H0887-20X100ML04061838172952
H0887-100ML04061838255921

Questions

1–4 of 4 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. Hello! Is it dissolved in H2O entirely? or does it have other surfactants?

    1 answer
    1. This product is comprised of HEPES free acid in tissue culture grade water. No other additives or preservatives are present, including surfactants.

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  4. Bonjour, Est-ce que ce produit H0887 peut-être congelé à -80°C sans risque de le faire précipiter ?

    1 answer
    1. It is best not to freeze the product as received. However, should the product be inadvertently frozen, it is best to thaw the product slowly. Do not place the frozen product in an incubator or water bath. Once thawed, inspect for any precipitates before use. A clear, colorless solution will be suitable for further use. Should this buffer be required to prepare the stock solution of another reagent that requires freezer storage, thaw slowly if possible.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-only review