Skip to Content
Merck
All Photos(2)

Key Documents

H0500

Sigma-Aldrich

1,2,3,4-Tetrahydroharmane-3-carboxylic acid

Synonym(s):

1-Methyl-1,2,3,4-Tetrahydropyrido[3,4-b]indole-3-carboxylic acid, 1-Methyl-1,2,3,4-tetrahydro-β-carboline-3-carboxylic acid, NSC 26225

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14N2O2
CAS Number:
Molecular Weight:
230.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

storage temp.

−20°C

Quality Level

SMILES string

CC1NC(Cc2c1[nH]c3ccccc23)C(O)=O

InChI

1S/C13H14N2O2/c1-7-12-9(6-11(14-7)13(16)17)8-4-2-3-5-10(8)15-12/h2-5,7,11,14-15H,6H2,1H3,(H,16,17)

InChI key

ZUPHXNBLQCSEIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in synthesis of molecules for biological studies including:
  • Cytotoxicity and insecticidal activities of harmine derivatives
  • Isoquinolines, β-carbolines, and 3-deazapurines via oxidative decarboxylation
  • Cytotoxic evaluation of 1,3-di- and 1,3,9-trisubstituted β-carbolines

Biochem/physiol Actions

Implicated in eosinophilia-myalgia syndrome associated with L-tryptophan ingestion.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Analysis of L-tryptophan for the etiology of eosinophilia-myalgia syndrome.
MMWR. Morbidity and mortality weekly report, 39(34), 589-591 (1990-08-31)
A comparative study of tissue distribution and excretion among three substances implicated in eosinophilia-myalgia syndrome.
J Adachi et al.
Advances in experimental medicine and biology, 398, 365-370 (1996-01-01)
S Fukushima et al.
Alcohol (Fayetteville, N.Y.), 9(1), 31-35 (1992-01-01)
We conducted analyses of 1-methyl-1,2,3,4-tetrahydro-beta-carboline-3-carboxylic acid (1Me3C-THBC) by gas chromatography-mass spectrometry (negative chemical ionization mode) to investigate its presence and the in vivo condensation between tryptophan and AcH. 1Me3C-THBC was found in the cerebellum and the cerebrum of normal rat
J Adachi et al.
Archives of toxicology, 67(4), 284-289 (1993-01-01)
Peak E substance, 1,1'-ethylidenebis[tryptophan], a contaminant found in L-tryptophan tablets, has been suggested as a causative agent for eosinophilia-myalgia syndrome (EMS). Peak E substance (50 mg/kg) was administered perorally to Wistar rats to determine its metabolism and distribution. A purification
Formation and determination of aldehyde-addition products of tryptophan.
K Meyer et al.
Advances in experimental medicine and biology, 398, 711-717 (1996-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service