Skip to Content
Merck
All Photos(1)

Key Documents

H0402

Sigma-Aldrich

Heparin−Agarose

(1:1 suspension in a 20% ethanol solution)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

biological source

heparin from Porcine intestinal mucosa

Quality Level

form

(1:1 suspension in a 20% ethanol solution)

matrix

4% beaded agarose

matrix activation

epichlorohydrin

matrix attachment

terminal aldehyde by reductive amination to amine linker

matrix spacer

7 atoms

capacity

≥0.5 mg/mL binding capacity (thrombin)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Heparin agarose is developed from porcine intestinal mucosa and is used in affinity chromatography. Heparin agarose has been used in studies to provide information on human monocytic ehrlichiosis, tumor necrosis and the effects of coagulation from Vipera snake venom.

Physical form

1:1 suspension in a 20% ethanol solution

Preparation Note

Prepared by end-point attachment for high-efficiency fractionation of antithrombin III and other specific binding proteins

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Zhou et al.
Journal of molecular biology, 271(3), 362-373 (1997-08-22)
Tn5 transposase (Tnp) binds to Tn5 and IS50 end inverted repeats, the outside end (OE) and the inside end (IE), to initiate transposition. We report the isolation of four Tnp mutants (YH41, TP47, EK54 and EV54) that increase the OE-mediated
Glycosaminoglycan binding assays.
A J Hoogewerf et al.
Methods in molecular biology (Clifton, N.J.), 138, 173-177 (2000-06-07)
Yongcheng Wang et al.
Molecular cell, 15(3), 343-353 (2004-08-12)
Amyloid beta-peptide, which forms neuronal and vascular amyloid deposits in Alzheimer's disease, is derived from an integral membrane protein precursor. The biological function of the precursor is currently unclear. Here we describe the X-ray structure of E2, the largest of
Kenji Kashiwagi et al.
Biomaterials, 30(6), 1166-1175 (2008-11-22)
Efficient immobilization of biomacromolecules on material surfaces is a key to development in areas of regenerative medicine and tissue engineering. However, strong and irreversible immobilization of cytokines on surfaces often diminishes their biological functionality. A destructive hydrophobic interaction between the
W H Yu et al.
The Journal of biological chemistry, 275(6), 4183-4191 (2000-02-08)
Many matrix metalloproteinases (MMPs) are tightly bound to tissues; matrilysin (MMP-7), although the smallest of the MMPs, is one of the most tightly bound. The most likely docking molecules for MMP-7 are heparan sulfate proteoglycans on or around epithelial cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service