GW20072F
Anti-C1QA antibody produced in chicken
affinity isolated antibody, buffered aqueous solution
Synonym(s):
Anti-Complement C1q subcomponent subunit A
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
Recommended Products
biological source
chicken
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
human
manufacturer/tradename
Genway 15-288-20072F
technique(s)
indirect ELISA: suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... C1QA(712)
Immunogen
Complement C1q subcomponent subunit A native protein.
Immunogen Sequence: GI # 7705753, sequence 1-245
Application
Anti-C1QA antibody produced in chicken is suitable for indirect ELISA and western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.
Biochem/physiol Actions
Complement C1q subcomponent subunit A is a protein encoded by the C1QA gene in humans. Anti-C1q antibodies are found to be pathogenic in the target organ damage of many autoimmune diseases, by facilitating C1q deposition and enhancing complement activation through classical pathway. It is also found to be associated with glomerular basement membrane (GBM) disease and in the pathogenesis of lupus. C1q deficiency leads to the development of autoimmune pathology throughout the life. C1q-binding protein participates in C1q-mediated chemotaxis of blood immune cells.
Physical form
Solution in phosphate buffered saline containing 0.02% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Hend Jlajla et al.
Immunobiology, 219(3), 241-246 (2013-12-18)
Hereditary C1q deficiency (C1qD) is the most penetrant genetic factor predisposing to the development of lupus pathology with more than 93% of C1q deficient patients developing this autoimmune pathology throughout their life. It is a rare autosomal recessive deficiency, with
B Namjou et al.
Lupus, 21(10), 1113-1118 (2012-04-05)
Homozygous C1q deficiency is an extremely rare condition and strongly associated with systemic lupus erythematosus. To assess and characterize C1q deficiency in an African-American lupus pedigree, C1q genomic region was evaluated in the lupus cases and family members. Genomic DNA
Shui-Yi Hu et al.
BMC immunology, 14, 42-42 (2013-09-24)
Anti-glomerular basement membrane (GBM) disease is a well-known antibody-induced autoimmune disease. A few patients have glomerular C1q deposition, but it is usually absent on renal histopathology. The role of C1q deposition in kidney injury is unclear. Recently, anti-C1q antibodies are
Muriel Tahtouh et al.
Journal of neuroinflammation, 9, 37-37 (2012-02-24)
In invertebrates, the medicinal leech is considered to be an interesting and appropriate model to study neuroimmune mechanisms. Indeed, this non-vertebrate animal can restore normal function of its central nervous system (CNS) after injury. Microglia accumulation at the damage site
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service