Skip to Content
Merck
All Photos(1)

Key Documents

GE17-5446-52

Ficoll®-Paque PREMIUM 1.073

Cytiva 17-5446-52, pack of 6 × 100 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
NACRES:
NA.25

form

liquid

packaging

pack of 6 × 100 mL

manufacturer/tradename

Cytiva 17-5446-52

color

Colorless to slightly yellow

pH range

5.5-7.5

General description

Ficoll-Paque PREMIUM products are a range of sterile, ready-to-use density gradient media for the preparation of mononuclear cells for use in cell therapy applications.

Application

Ficoll-Paque PREMIUM products are based on Ficoll-Paque PLUS, which has been used for more than 30 years as a sterile density medium for the isolation of high yields of mononuclear cells from bone marrow, peripheral blood, and cord blood. All Ficoll-Paque PREMIUM products differ from Ficoll-Paque PLUS in that they are manufactured under a Quality Management System certified to ISO 13485:2003.

Features and Benefits

  • Sterile, ready to use reagents used to prepare mononuclear cell preparations from peripheral blood, bone marrow, and umbilical cord blood by density gradient centrifugation.
  • Manufactured under a Quality Management System certified to ISO 13485:2003.
  • Low levels of endotoxin (< 0.12 EU/mL) secured and tested.
  • Cytotoxicity grade 0.
  • A Regulatory Support File (RSF) is available for Ficoll-Paque PREMIUM products.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C ()

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Ficoll is a registered trademark of Cytiva

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Density Marker Beads facilitate Percoll gradient monitoring in biological separations.

This page shows methods to remove Percoll from a sample after centrifugation.

Density Marker Beads facilitate Percoll gradient monitoring in biological separations.

Density Marker Beads facilitate Percoll gradient monitoring in biological separations.

See All

Protocols

The following procedure has been evaluated with Ficoll-Paque PLUS and is recommended for separation of normal blood samples for maximum reproducibility.

Related Content

Data was compiled to assist the researcher in selecting references most likely to contain relevant information regarding use of Percoll for a particular cell or tissue type.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service