Skip to Content
Merck

G9768

Guanosine 5′-triphosphate–Agarose

aqueous glycerol suspension

Synonym(s):

5′-GTP agarose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106500
NACRES:
NA.56
MDL number:
Biological source:
bacterial (CORYNEBACTERIUM SP), enzyme from Porcine brain, plant (Sea weed), yeast
Form:
aqueous glycerol suspension
Storage temp.:
−20°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

biological source

bacterial (CORYNEBACTERIUM SP), enzyme from Porcine brain, plant (Sea weed), yeast

form

aqueous glycerol suspension

extent of labeling

1-5 μmol per mL

matrix

cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

ribose hydroxyls

matrix spacer

11 atoms (adipic acid dihydrazide)

storage temp.

−20°C

SMILES string

[X]O[C@H]1[C@@H](O[C@@H]([C@H]1O[X])CO[P](=O)(O[P](=O)(O[P](=O)(O)O)O)O)[n]2c3nc(nc(c3nc2)O)N

Application

Guanosine 5′-triphosphate-agarose (5′-GTP agarose) is an agarose conjugate in aqueous glycerol suspension used in affinity chromatography, for purification of guanosine-binding proteis

Physical form

Suspension in 50% glycerol containing 0.25 M NaCl
Guanosine 5′-triphosphate–Agarose has been used:
  • in chromatographic column for the purification of a translocation stimulating factor
  • in GTP-Agarose pull down assays to measure the levels of GTP-bound RHEB (Ras homolog enriched in brain)
  • in GTP-Binding assay
  • for the depletion of GTP binding proteins in cytosol of rat liver


Storage Class

10 - Combustible liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library