Skip to Content
Merck
All Photos(2)

Key Documents

G9297

Sigma-Aldrich

Glutathione Reductase human

buffered aqueous solution, ≥10 units/mg protein, recombinant, expressed in E. coli

Synonym(s):

GR, Glutathione-disulfide reductase, NADPH:oxidized glutathione oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

recombinant

expressed in E. coli

Quality Level

form

buffered aqueous solution

specific activity

≥10 units/mg protein

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GSR(2936)

General description

Research area: Cell signaling. Glutathione Reductase belongs to the homodimericFAD−disulfide oxidoreductases family. is made up of highly conserved domains such as two Rossmann fold domains, where one binds to reduced nicotinamide adenine dinucleotide phosphate (NADPH) and flavin adenine dinucleotide (FAD) the second one is an interface dimerization domain. It favors its accumulation in the regions of high electron flux in cells where reactive species are generated.[1]

Application

Human glutathione reductase has been used in:
  • glutathione reductase activity assay[2]
  • oxidative stress analysis[3]
  • redox assays[4]

Biochem/physiol Actions

Glutathione reductase enzyme is a homodimeric enzyme containing 1 FAD molecule and 1 NADPH binding domain per subunit. Both human GR (hGR) and Plasmodium falciparum GR (PfGR) are essential for the survival of the malaria parasite within the human erythrocyte. Thus, this enzyme may be used for studies of candidate anti-malaria reagents.
Glutathione reductase is a ubiquitous flavoenzyme involved in the protection from cell stress. Glutathione reductase catalyzes the reduction of oxidized glutathione (GSSG) to glutathione (GSH). It is essential for the glutathione redox cycle that maintains adequate levels of reduced cellular GSH, which serves as an antioxidant reacting with free radicals and organic peroxides. Glutathione is also an electron donor for glutathione peroxidases and a substrate for glutathione S-transferases contributing to the detoxification and elimination of toxic electrophilic metabolites and xenobiotics.
Human glutathione reductase is suitable as cytostatic and antimalarialagent. It may also be used to protect against malaria by mimicking favism by blockingthe enzyme with specific inhibitors.

Unit Definition

1 unit will reduce 1.0 μmole of DTNB to TNB per minute at 25 °C at pH 7.5.

Physical form

Solution containing 25 mM Tris-HCl, pH 7.4, 1 mM EDTA, and 50% (v/v) glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dietary methionine can sustain cytosolic redox homeostasis in the mouse liver
Sofi Eriksson, et al.
Nature Communications (2014)
Definition of transcriptome-based indices for quantitative characterization of chemically disturbed stem cell development: introduction of the STOP-Toxukn and STOP-Toxukk tests.
Vaibhav Shinde, et al.
Archives of Toxicology, 91(2) (2017)
Haitao Shi et al.
Plant physiology and biochemistry : PPB, 71, 226-234 (2013-08-27)
As a gaseous molecule, hydrogen sulfide (H2S) has been recently found to be involved in plant responses to multiple abiotic stress. In this study, salt (150 and 300 mM NaCl), osmotic (15% and 30% PEG6000) and cold (4 °C) stress
C C Bohme et al.
The Journal of biological chemistry, 275(48), 37317-37323 (2000-09-02)
The homodimeric flavoenzyme glutathione reductase (GR) maintains high intracellular concentrations of the antioxidant glutathione (GSSG + NADPH + H(+) <--> 2 GSH + NADP(+)). Due to its central function in cellular redox metabolism, inhibition of GR from the malarial parasite
Walid Abu El-Soud et al.
Plant physiology and biochemistry : PPB, 71, 173-183 (2013-08-14)
Seed germination and growth of seedlings are critical phases of plant life that are adversely affected by various environmental cues. Water availability is one of the main factors that limit the productivity of many crops. This study was conducted to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service