Skip to Content
Merck

G8673

β-Glucanase 2, thermostable

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

β-Glucanase, thermostable

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
MDL number:
NACRES:
NA.54
UNSPSC Code:
12352204
Specific activity:
≥1.0 units/mg protein
Assay:
≥90% (SDS-PAGE)
Recombinant:
expressed in E. coli
Concentration:
≥20 mg protein/mL (Bradford)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in E. coli

assay

≥90% (SDS-PAGE)

form

liquid

specific activity

≥1.0 units/mg protein

mol wt

38 kDa

concentration

≥20 mg protein/mL (Bradford)

shipped in

wet ice

storage temp.

2-8°C

Application

β-Glucanase is used to study cell wall modifications and for carbohydrate hydrolysis. It has been used to supplement barley-based diets for poultry and to study the reopening signal conduits and release of dormancy in the Populus species.

Biochem/physiol Actions

β-Glucanase is involved in cell wall modifications and carbohydrate hydrolysis.

Physical form

Supplied as a solution in 50 mM Tris-HCl, pH 7.5, 100 mM NaCl, and 25% glycerol.

Other Notes

One unit will produce 1 μmole of reducing sugar (measured as glucose) from Beta-glucan per minute at pH 5.8 at 70 °C.


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library