Skip to Content
Merck
All Photos(2)

Documents

G7550

Sigma-Aldrich

Sephadex® G-75

Superfine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

technique(s)

buffer exchange: suitable

matrix active group

phase

swelling

1 g swells to 12-15 mL

bead size

20-50 μm

application(s)

life science and biopharma

compatibility

Cytiva

Looking for similar products? Visit Product Comparison Guide

General description

Sephadex® G-75 is well-established gel filtration medium for desalting and buffer exchange of large biomolecules >80,000 molecular weight.
Sephadex® is a gel filtration medium prepared by crosslinking dextran with epichlorohydrin. Different types of Sephadex differ in their degree of cross-linking and hence in their degree of swelling and their molecular fractionation range. Sephadex® G-75 is for larger molecules.

Application

Sephadex® G-75 is a gel filtration media used in gel filtration chromatography and protein chromatography. It has been used for desaltation process to study the production, purification, and immobilization of l-asparaginase II (ASNase II) in chitosan nanoparticles (CSNPs).

Legal Information

Sephadex is a registered trademark of Cytiva
Sepharose is a trademark of Cytiva

replaced by

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Engineering a Disulfide Bond in Recombinant Manganese Peroxidase Results in Increased Thermostability
Reading, N.S., and Aust, S.D.
Biotechnol. Progress, 16(3), 326-333 (2000)
Sofia R Pauleta et al.
Biochemistry, 43(35), 11214-11225 (2004-09-16)
The gene for pseudoazurin was isolated from Paracoccus pantotrophus LMD 52.44 and expressed in a heterologous system with a yield of 54.3 mg of pure protein per liter of culture. The gene and protein were shown to be identical to
John G Vontas et al.
The Biochemical journal, 362(Pt 2), 329-337 (2002-02-21)
A novel glutathione S-transferase (GST)-based pyrethroid resistance mechanism was recently identified in Nilaparvata lugens [Vontas, Small and Hemingway (2001) Biochem. J. 357, 65-72]. To determine the nature of GSTs involved in conferring this resistance, the GSTs from resistant and susceptible
L Shilpa Satheesh et al.
Indian journal of experimental biology, 49(5), 366-374 (2011-05-28)
Antimicrobial activity of protease inhibitor isolated from Coccinia grandis (L.) Voigt. has been reported. A 14.3 kDa protease inhibitor (PI) was isolated and purified to homogeneity by ammonium sulfate precipitation (20-85% saturation), sephadex G-75, DEAE sepharose column and trypsin-sepharose affinity
Fakhra Amin et al.
Protein and peptide letters, 18(2), 210-218 (2010-11-09)
Cystatins are thiol proteinase inhibitors ubiquitously present in mammalian body and serve various important physiological functions. To purify and characterize Thiol protease inhibitor from buffalo brain and to compare its properties with respect to tissue and organ difference from other

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service