Skip to Content
Merck
All Photos(1)

Key Documents

G6910

Supelco

Ginkgolide B from Ginkgo biloba leaves

≥90% (HPLC)

Synonym(s):

BN-52021

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O10
CAS Number:
Molecular Weight:
424.40
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

Quality Level

Assay

≥90% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

C[C@@H]1C(=O)O[C@H]2[C@H](O)[C@@]34[C@H]5C[C@@H](C(C)(C)C)[C@@]36[C@@H](O)C(=O)O[C@H]6O[C@@]4(C(=O)O5)[C@@]12O

InChI

1S/C20H24O10/c1-6-12(23)28-11-9(21)18-8-5-7(16(2,3)4)17(18)10(22)13(24)29-15(17)30-20(18,14(25)27-8)19(6,11)26/h6-11,15,21-22,26H,5H2,1-4H3/t6-,7+,8-,9+,10+,11+,15+,17+,18+,19-,20-/m1/s1

InChI key

SQOJOAFXDQDRGF-MMQTXUMRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ginkgolide B from Ginkgo biloba leaves is a terpene lactone and one of the main molecular form of ginkgolides, present in Ginkgo biloba leaves. It is widely used to treat peripheral and cerebral circulation disorders.[1]

Application

Ginkgolide B from Ginkgo biloba leaves is used as an analytical reference standard:
  • For the chromatographic separation of the analyte in Ginkgo biloba leaves extract and to study the antimicrobial activity exerted on a variety of Gram-positive and -negative bacteria and yeasts.[2]
  • For the determination of the analyte in Ginkgo biloba L. phytopharmaceuticals using high-performance liquid chromatography technique.[1]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Primary constituent of Ginkgo biloba. Potential anticancer agent and neuroprotective.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antimicrobial investigation of semi-purified fractions of Ginkgo biloba leaves
Mazzanti.G, et al.
Journal of Ethnopharmacology, 71(1-2), 83-88 (2005)
Yuna Cui et al.
Fitoterapia, 83(5), 913-920 (2012-04-28)
To screen the presence of ginkgolide B-producing endophytic fungi from the root bark of Ginkgo biloba, a total of 27 fungal isolates, belonging to 6 different genus, were isolated from the internal root bark of the plant Ginkgo biloba. The
Shi-hai Xia et al.
Chinese medical journal, 120(10), 922-928 (2007-06-05)
To review the recent research progress in pharmacological actions and mechanisms of ginkgolide B. Data sources Information included in this article was identified by searching of PUBMED (1987 - 2006) online resources using the key terms "ginkgolide B", "platelet activating
Peng Lv et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 44(3), 235-240 (2011-08-23)
Cerebral ischemia and reperfusion is one of the leading causes for death and severe disabilities in the world and often lead to irreversible brain damage over later lifespan. The aim of this study was to investigate the evolution of pathological
Karyn M Maclennan et al.
Progress in neurobiology, 67(3), 235-257 (2002-08-10)
Ginkgo biloba extracts such as EGb-761 have been suggested to have a multitude of beneficial effects on CNS function, from enhancing cognitive function in dementia to facilitating recovery from acute forms of neural damage such as hypoxia/ischemia. Ginkgolide B, one

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service