Skip to Content
Merck

G6513

β-D-Glucan from barley

≥95% (HPLC)

Synonym(s):

beta-Glucan

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

plant (barley)

Quality Segment

assay

≥95% (HPLC)

form

powder

color

white to faint yellow

storage temp.

2-8°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)OC2[C@H](O[C@H]([C@@H]([C@H]2O)O)OC3[C@H](O[C@@H]([C@@H]([C@H]3O)O)O)CO)CO

InChI

1S/C18H32O16/c19-1-4-7(22)8(23)12(27)17(31-4)34-15-6(3-21)32-18(13(28)10(15)25)33-14-5(2-20)30-16(29)11(26)9(14)24/h4-29H,1-3H2/t4-,5-,6-,7-,8+,9-,10-,11-,12-,13-,14?,15?,16+,17+,18+/m1/s1

InChI key

FYGDTMLNYKFZSV-URKRLVJHSA-N

Biochem/physiol Actions

β-D-Glucan is a biomarker used for the fungal infection, candidiasis. In a clinical study β-D-glucan levels were found to have possible usefulness in the prediction of patient outcomes.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Still not finding the right product?

Explore all of our products under β-D-Glucan from barley


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library