Skip to Content
Merck

G5376

DL-Glyceraldehyde 3-phosphate diethyl acetal barium salt

Synonym(s):

3,3-Diethoxy-1,2-propanediol 1-phosphate barium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7H15BaO7P
CAS Number:
Molecular Weight:
379.49
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
EC Number:
300-986-5
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

impurities

<2.0% water (Karl Fischer)

color

white

cation traces

Ba: 28.8-42.7% (anhydrous)

storage temp.

−20°C

SMILES string

[Ba++].CCOC(OCC)C(O)COP([O-])([O-])=O

InChI

1S/C7H17O7P.Ba/c1-3-12-7(13-4-2)6(8)5-14-15(9,10)11;/h6-8H,3-5H2,1-2H3,(H2,9,10,11);/q;+2/p-2

InChI key

OCALAXUAYOULKA-UHFFFAOYSA-L

Biochem/physiol Actions

DL-Glyceraldehyde 3-phosphate has been shown to function as an inhibitor of aspartate aminotransferase isozymes.

Packaging

A package of specially washed Dowex 50× 4-200R is also included for preparing a solution of glyceraldehyde 3-phosphate from this derivative at no extra charge. Instructions for preparing this solution from this derivative accompanies each package.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information



pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library