Skip to Content
Merck
All Photos(1)

Key Documents

G5045

Sigma-Aldrich

GW6471

≥98% (HPLC), powder, PPARα antagonist

Synonym(s):

N-((2S)-2-(((1Z)-1-Methyl-3-oxo-3-(4-(trifluoromethyl)phenyl)prop-1-enyl)amino)-3-(4-(2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)ethoxy)phenyl)propyl)propanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H36F3N3O4
CAS Number:
Molecular Weight:
619.67
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

GW6471, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

CCC(=O)NC[C@H](Cc1ccc(OCCc2nc(oc2C)-c3ccccc3)cc1)N\C(C)=C/C(=O)c4ccc(cc4)C(F)(F)F

InChI

1S/C35H36F3N3O4/c1-4-33(43)39-22-29(40-23(2)20-32(42)26-12-14-28(15-13-26)35(36,37)38)21-25-10-16-30(17-11-25)44-19-18-31-24(3)45-34(41-31)27-8-6-5-7-9-27/h5-17,20,29,40H,4,18-19,21-22H2,1-3H3,(H,39,43)/b23-20-/t29-/m0/s1

InChI key

TYEFSRMOUXWTDN-DYQICHDWSA-N

General description

GW6471 prevents apoptosis and cell cycle arrest at G0/G1 in renal cell carcinoma. It reduces enhanced fatty acid oxidation and oxidative phosphorylation associated with glycolysis inhibition.

Application

GW6471 has been used:
  • to investigate the role of peroxisome proliferator-activated receptors (PPARs) alpha in perfluorooctanoic acid (PFOA) induced developmental cardiotoxicity
  • to abolish the protection of bezafibrate (BZA) against hypertrophy
  • to block the enhanced activity of the hepatitis B virus (HBV) core promoter

Biochem/physiol Actions

GW6471 is a PPARα antagonist, which completely inhibits GW409544-induced activation of PPARα with an IC50 = 0.24 μM. GW6471 induces a PPARα conformation that interacts efficiently with co-repressors.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Nuclear Receptors (PPARs) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PPARalpha inhibition modulates multiple reprogrammed metabolic pathways in kidney cancer and attenuates tumor growth
Abu AO, et al.
American Journal of Physiology. Cell Physiology, 308(11), C890-C898 (2015)
Shiori Ikeguchi et al.
Journal of pharmacological sciences, 138(1), 9-15 (2018-09-24)
10-oxo-trans-11-octadecenoic acid (KetoC) and 10-hydroxy-cis-12-octadecenoic acid (HYA) are long-chain fatty acids generated from linoleic acid by the gut lactic acid bacterium Lactobacillus plantarum. These fatty acids have been reported to have anti-inflammatory activity in the intestine. However, little is known
Hongliang Li et al.
European journal of pharmacology, 812, 155-162 (2017-07-19)
We examined the effects of the PPARα activator fenofibrate on voltage-dependent K
Chun Wang et al.
Environmental health perspectives, 119(12), 1739-1744 (2011-08-19)
Numerous man-made pollutants activate the aryl hydrocarbon receptor (AhR) and are risk factors for type 2 diabetes. AhR signaling also affects molecular clock genes to influence glucose metabolism. We investigated mechanisms by which AhR activation affects glucose metabolism. Glucose tolerance
Bezafibrate attenuates pressure overload-induced cardiac hypertrophy and fibrosis
Xu SC, et al.
PPAR Research, 2017 (2017)

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service