Skip to Content
Merck
All Photos(2)

Key Documents

G1163

Sigma-Aldrich

O-Glycosidase from Streptococcus pneumoniae

recombinant, expressed in E. coli, buffered aqueous solution

Synonym(s):

Endo-α-N-acetylgalactosaminidase, O-Glycanase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

recombinant

expressed in E. coli

Quality Level

conjugate

(O-linked)

form

buffered aqueous solution

mol wt

180 kDa

concentration

≥800 units/mL

shipped in

wet ice

storage temp.

2-8°C

Biochem/physiol Actions

Releases unsubstituted Ser- and Thr-linked β-Gal-(1→3)-α-GalNAc (Core 1 type O-glycan) from glycoproteins. Substitutions of the disaccharide core with sialic acid, lactosamine (galactose-N-acetyl glucosamine), or fucose will block hydrolysis and prevent the liberation of the oligosaccharide from the protein. Pretreament with glycolytic enzymes to remove substituent saccharides from the O-glycan may be needed prior to cleavage using O-glycosidase..

Packaging

Supplied with 5× Reaction Buffer, 250 mM NaH2PO4 pH 5.0.

Unit Definition

One unit will hydrolyze 1 μmole of p-nitrophenyl galacto-N-bioside (β-Gal-(1→3)-α-GalNAc-1→ΟC6H4NO2) per min at 37 °C at pH 6.5.

Physical form

Solution in 50 mM sodium phosphate, pH 7.5

Analysis Note

Screened for presence of: β-galactosidase, α-mannosidase, β-hexosaminidase, α-fucosidase, neuraminidase, and proteases. See Certificate of Analysis for lot specific information.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Brockhausen
Biochimica et biophysica acta, 1473(1), 67-95 (1999-12-02)
Glycoproteins with O-glycosidically linked carbohydrate chains of complex structures and functions are found in secretions and on the cell surfaces of cancer cells. The structures of O-glycans are often unusual or abnormal in cancer, and greatly contribute to the phenotype
Nelia A Tobey et al.
Digestive diseases and sciences, 55(7), 1856-1865 (2010-05-27)
The structures that contribute to shunt resistance (Rs) in esophageal epithelium are incompletely understood, with 35-40% of Rs known to be calcium-dependent, reflecting the role of e-cadherin. Two calcium-independent candidates for the remaining approximately 60% of Rs have been identified:
Roudabeh J Jamasbi et al.
Hybridoma and hybridomics, 22(6), 367-376 (2003-12-20)
A mouse monoclonal antibody (MAb-9) produced by immunization with a human esophageal carcinoma cell line, TE-2 (derived from undifferentiated squamous cell carcinoma) reacted specifically with about 30% of esophageal carcinoma cell lines and tissue sections from clinical samples. MAb-9 showed
K Sonoda et al.
Cancer, 77(8), 1501-1509 (1996-04-15)
A large number of monoclonal antibodies (MoAbs) against human tumor cells have been generated and it has been shown that these MoAbs are useful tools in the diagnosis and treatment of cancer patients, as well as in the basic investigation
H Yamini Shrivastava et al.
Journal of biomolecular structure & dynamics, 21(5), 671-680 (2004-02-11)
In the present study, the impact of chromium(III) complexes ([Cr(salen)(H2O)2](+) (1), [Cr(en)3]3+ (2) and [Cr(EDTA)(H2O)]- (3)) on the biophysical properties of mucin like specific viscosity, zeta potential and particle size has been investigated. It is evident from the present investigation

Articles

Learn about O-linked glycan strategies, O-glycosidase actions, how to remove sialic acid residues, β-Elimination, and O-glycan modifications.

Learn about O-linked glycan strategies, O-glycosidase actions, how to remove sialic acid residues, β-Elimination, and O-glycan modifications.

Learn about O-linked glycan strategies, O-glycosidase actions, how to remove sialic acid residues, β-Elimination, and O-glycan modifications.

Learn about O-linked glycan strategies, O-glycosidase actions, how to remove sialic acid residues, β-Elimination, and O-glycan modifications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service