Skip to Content
Merck

G0141

L-Glutamic acid γ-(4-methoxy-β-naphthylamide)

≥98% (TLC), powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H18N2O4
CAS Number:
Molecular Weight:
302.33
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

L-Glutamic acid γ-(4-methoxy-β-naphthylamide),

assay

≥98% (TLC)

Quality Segment

form

powder

solubility

1 N HCl: ethanol, 1:1: 25 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

OC([C@@H](N)CCC(NC1=CC2=CC=CC=C2C(OC)=C1)=O)=O

InChI

1S/C16H18N2O4/c1-22-14-9-11(8-10-4-2-3-5-12(10)14)18-15(19)7-6-13(17)16(20)21/h2-5,8-9,13H,6-7,17H2,1H3,(H,18,19)(H,20,21)

InChI key

DUNDTSUHZJULAX-UHFFFAOYSA-N

Application

L-Glutamic acid γ-(4-methoxy-β-naphthylamide) has been used in histochemical staining of HepG2 cells for γ-glutamyl transpeptidase (GGT) subcellular localization.

Biochem/physiol Actions

L-Glutamic acid γ-(4-methoxy-β-naphthylamide) or GMNA is a synthetic substrate of γ-glutamyl transpeptidase (GGT).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


pictograms

Health hazard

signalword

Warning

hcodes

pcodes

Hazard Classifications

Carc. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library