FLAAB
Adenosine 5′-triphosphate (ATP) assay mix dilution buffer
lyophilized powder
Synonym(s):
ATP Assay Dilution Buffer, Assay Mix Buffer, Assay Mix for ATP
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
eCl@ss:
32160414
NACRES:
NA.84
Recommended Products
General description
Adenosine 5′-triphosphate (ATP) assay mix dilution buffer is a component of the ATP bioluminescent assay kit that may be employed for the quantitative bioluminescent determination of ATP in experimental samples.
Application
Adenosine 5′-triphosphate (ATP) assay mix dilution buffer has been used to determine the level of adenosine kinase activity in isolated sections of mouse hippocampi. It has also been used to measure the ATP content in oocytes and harderian gland(HG) tissue homogenates.
Components
Adenosine 5′-triphosphate (ATP) assay mix dilution buffer contains MgSO4, dithiothreitol (DTT), ethylene diamine tetraacetic acid (EDTA, bovine serum albumin (BSA), and tricine buffer salts.
Reconstitution
Reconstitute with 50 mL of sterile water.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Peter Stief et al.
BMC microbiology, 14, 35-35 (2014-02-13)
A wealth of microbial eukaryotes is adapted to life in oxygen-deficient marine environments. Evidence is accumulating that some of these eukaryotes survive anoxia by employing dissimilatory nitrate reduction, a strategy that otherwise is widespread in prokaryotes. Here, we report on
Nicolette Gouder et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 24(3), 692-701 (2004-01-23)
Endogenous adenosine in the brain is thought to prevent the development and spread of seizures via a tonic anticonvulsant effect. Brain levels of adenosine are primarily regulated by the activity of adenosine kinase. To establish a link between adenosine kinase
Hana M Russo et al.
Journal of immunology (Baltimore, Md. : 1950), 197(4), 1353-1367 (2016-07-08)
Canonical inflammasome activation induces a caspase-1/gasdermin D (Gsdmd)-dependent lytic cell death called pyroptosis that promotes antimicrobial host defense but may contribute to sepsis. The nature of the caspase-1-dependent change in plasma membrane (PM) permeability during pyroptotic progression remains incompletely defined.
Andrea Boyd-Tressler et al.
The Journal of biological chemistry, 289(39), 27246-27263 (2014-08-13)
Anti-tumor immune responses have been linked to the regulated release of ATP from apoptotic cancer cells to engage P2 purinergic receptor signaling cascades in nearby leukocytes. We used the Jurkat T cell acute lymphocytic leukemia model to characterize the role
Nobuhiro Moro et al.
Experimental and therapeutic medicine, 21(6), 575-575 (2021-04-15)
The aim of the current study was to determine effects of mild traumatic brain injury (TBI), with or without blockade of purinergic ATP Y1 (P2Y1) receptors or store-operated calcium channels, on extracellular levels of ATP, glutamate, glucose and lactate. Concentrations
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service