Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352207
NACRES:
NA.71
Product Name
Medium 199 Earle′s with stable Glutamine, With Earle′s salts, stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Quality Segment
sterility
sterile-filtered
form
liquid
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, tested
components
stable glutamine: yes
Earle’s salts (5% CO2): yes
sodium pyruvate: no
NaHCO3: yes
phenol red: yes
HEPES: no
shipped in
ambient
storage temp.
2-8°C
General description
Complete liquid version of the original M 199 formulation published by Morgan.
Application
Originally developed as a completely defined media formulation for the culture of primary explants. These media have broad applicability, particularly for non-transformed cells and are widely used for vaccine production, primary pancreatic explants, and lens tissues.
Still not finding the right product?
Explore all of our products under Medium 199 Earle′s with stable Glutamine
Storage Class
13 - Non Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.