Skip to Content
Merck
All Photos(1)

Documents

F9636

Sigma-Aldrich

IgG−FITC from human serum

~20 mg/mL, buffered aqueous solution

Synonym(s):

IgG-Fluorescein isothiocyanate from human serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

conjugate

FITC conjugate

form

buffered aqueous solution

concentration

~20 mg/mL

storage temp.

2-8°C

target post-translational modification

unmodified

General description

IgGs are glycoprotein antibodies that regulate immunological defense mechanisms such as phagocytosis and inflammatory responses . IgG also modulates complement-mediated cell lysis . IgG-FITC from human serum does not react with purified human IgA, IgG, IgM, and Bence Jones κ and lambda myeloma proteins. Additionally, the product does not react with serum from rat, goat, rabbit, guinea pig, and mouse.

Application

IgG-FITC from human serum is suitable for use in immunoaffinity purifications , and metal-enhanced fluorescence . The product may also be used for immunoelectrophoretic applications.

Linkage

Do not confuse with Anti-Human IgG–FITC conjugate.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takeshi Hori et al.
Nature communications, 15(1), 962-962 (2024-02-09)
Human placental villi have essential roles in producing hormones, mediating nutrient and waste exchange, and protecting the fetus from exposure to xenobiotics. Human trophoblast organoids that recapitulate the structure of villi could provide an important in vitro tool to understand
Purification of immunoglobulins from Serum Using Thiophilic Cellulose Beads
Subramanian, A.
International Journal of Biochromatography, 5(I), 31-47 (2000)
Vishakha Sabharwal et al.
Infection and immunity, 77(3), 1121-1127 (2009-01-14)
Strategies to limit complement deposition on Streptococcus pneumoniae are established as virulence features for invasive disease, but their role in respiratory tract infection requires further analysis. We evaluated complement C3 protein deposition on discordant S. pneumoniae isolates of the same
Tatsuro Endo et al.
Biotechnology journal, 11(6), 831-837 (2016-05-07)
The development of high-sensitive, and cost-effective novel biosensors have been strongly desired for future medical diagnostics. To develop novel biosensor, the authors focused on the specific optical characteristics of photonic crystal. In this study, a label-free optical biosensor, polymer-based two-dimensional
C I Bindon et al.
The Journal of experimental medicine, 168(1), 127-142 (1988-07-01)
Humanized antibodies are likely to have a major role in therapy and it is important to define their interaction with physiological effectors. By comparing a matched series of chimeric human mAbs we found that igG1 was most efficient in complement

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service