Skip to Content
Merck
All Photos(4)

Key Documents

F9302

Sigma-Aldrich

Factor X Activated (Xa) from bovine plasma

aqueous glycerol solution

Synonym(s):

Factor Xa

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

form

aqueous glycerol solution

Quality Level

specific activity

≥50 units/mg protein

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

cow ... F10(280787)

Looking for similar products? Visit Product Comparison Guide

Application

Bovine Factor Xa is used for site specific cleavage of recombinant fusion proteins containing an accessible Factor Xa recognition site for removal of affinity tags.
Fusion proteins are commonly expressed with a factor Xa cleavable Ile-Glu (or Asp)-Gly-Arg-↓-X sequence. Typically 1 mg of fusion protein can be incubated with 10 μg of factor Xa for 2.5 hours at 37 °C.

Biochem/physiol Actions

Factor Xa catalyzes the hydrolysis of the Arg-Thr and then Arg-Ile bonds in prothrombin to yield active thrombin.
The fairly strict recognition sequence is Ile-Glu (or Asp)-Gly-Arg-↓-X.
It may sometimes cleave at other basic residues, depending on the conformation of the target protein. Factor Xa will not cleave if a proline residue follows the arginine of the recognition sequence.
pH Optimum: 7.6-8.0
Temperature Optimum: 37 °C

Physical properties

Factor Xa is a serine endoproteinase and a member of the S1 peptidase family. Factor Xa plays a critical role in the coagulation cascade by catalyzing the proteolytic conversion of prothrombin to active thrombin. Factor Xa′s prothrombin conversion activity is greatly enhanced in vivo when complexed with factor V, calcium ions and phospholipids on the activated platelet surface.
The zymogen form, Factor X, is activated in vivo by two different pathways. The intrinsic pathway utilizes a catalytic complex composed of factor IXa, factor VIII, phospholipids and calcium ions. The extrinsic pathway utilizes a complex of factor VII and tissue factor. The factor X zymogen is a 55 KDa glycoprotein with a light and heavy chain joined by a single disulfide.

Unit Definition

One unit of activated Factor X will liberate 1.0 μmole of p-nitroanilide from N-benzoyl-L-isoleucyl-L-glutamyl-L-glycyl-L-arginine-p-nitroaniline per minute at pH 8.3 at 37 °C.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

substrate

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genmin Lu et al.
Nature medicine, 19(4), 446-451 (2013-03-05)
Inhibitors of coagulation factor Xa (fXa) have emerged as a new class of antithrombotics but lack effective antidotes for patients experiencing serious bleeding. We designed and expressed a modified form of fXa as an antidote for fXa inhibitors. This recombinant
Antidotes edge closer to reversing effects of new blood thinners.
Elie Dolgin
Nature medicine, 19(3), 251-251 (2013-03-08)
Taki Galanis et al.
Hospital practice (1995), 41(1), 26-36 (2013-03-08)
Nonvalvular atrial fibrillation (AF) is the most common clinically significant cardiac arrhythmia occurring in patients in the United States. The primary clinical consequence of AF is an increase in the risk and severity of strokes. Treatment guidelines recommend anticoagulation therapy
Alexander T Cohen et al.
The New England journal of medicine, 368(6), 513-523 (2013-02-08)
The clinically appropriate duration of thromboprophylaxis in hospitalized patients with acute medical illnesses is unknown. In this multicenter, randomized, double-blind trial, we evaluated the efficacy and safety of oral rivaroxaban administered for an extended period, as compared with subcutaneous enoxaparin
[Xarelto: other advantages].
Perspective infirmiere : revue officielle de l'Ordre des infirmieres et infirmiers du Quebec, 10(1), 59-59 (2013-02-06)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service