Skip to Content
Merck

F9272

α-1,2-Fucosidase solution

buffered aqueous solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
EC Number:
232-914-2
NACRES:
NA.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

buffered aqueous solution

Quality Segment

specific activity

≥0.4 units/mg protein

packaging

vial of 0.004 unit

UniProt accession no.

foreign activity

β-galactosidase, α-mannosidase, β-hexosaminidase, neuraminidase, α-1-3,4-fucosidase and proteases, none detected

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... FUCA1(2517)

Biochem/physiol Actions

Releases α-1,2-fucose from the non-reducing end of complex carbohydrates.

Physical form

Solution in 20 mM Tris-HCl, pH 7.5, 25 mM NaCl

Other Notes

One unit will release 1.0 μmole of fucose from 2′-fucosyllactose per min at pH 5.0 at 37°C


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library