Skip to Content
Merck

F8764

Fast Red TR Salt hemi(zinc chloride) salt

Synonym(s):

4-Chloro-2-methylbenzenediazonium salt, Azoic Diazo No. 11

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7H6Cl2N2 · 0.5 ZnCl2
CAS Number:
Molecular Weight:
257.19
UNSPSC Code:
12352204
NACRES:
NA.83
PubChem Substance ID:
MDL number:
Colour Index Number:
37085
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

composition

Dye content, >90%

solubility

water: 25 mg/mL, clear, colorless to light yellow

storage temp.

−20°C

SMILES string

[Cl-].[Cl-].Cl[Zn]Cl.Cc1cc(Cl)ccc1[N+]#N.Cc2cc(Cl)ccc2[N+]#N

InChI

1S/2C7H6ClN2.4ClH.Zn/c2*1-5-4-6(8)2-3-7(5)10-9;;;;;/h2*2-4H,1H3;4*1H;/q2*+1;;;;;+2/p-4

InChI key

RIIVFDKBXXMYRL-UHFFFAOYSA-J



signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Corr. 1C

Storage Class

4.1A - Other explosive hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library