Skip to Content

F8758

Folic acid

BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%

Synonym(s):

Pterylmonoglutamic acid, PteGlu, Pteroyl-L-glutamic acid, Vitamin M

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H19N7O6
CAS Number:
Molecular Weight:
441.40
NACRES:
NA.75
PubChem Substance ID:
eCl@ss:
34058014
UNSPSC Code:
12352205
EC Number:
200-419-0
MDL number:
Beilstein/REAXYS Number:
100781
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

product line

BioReagent

assay

≥97%

form

powder

mol wt

Mw 441.40 g/mol

technique(s)

cell culture | insect: suitable, cell culture | mammalian: suitable, cell culture | plant: suitable

color

dark yellow

solubility

1 M NaOH: soluble 50 mg/mL, clear to very slightly hazy, faintly yellow to dark yellow-orange

storage temp.

room temp

SMILES string

NC(N1)=NC(C2=C1N=CC(CNC3=CC=C(C(N[C@@H](CCC(O)=O)C(O)=O)=O)C=C3)=N2)=O

InChI

1S/C19H19N7O6/c20-19-25-15-14(17(30)26-19)23-11(8-22-15)7-21-10-3-1-9(2-4-10)16(29)24-12(18(31)32)5-6-13(27)28/h1-4,8,12,21H,5-7H2,(H,24,29)(H,27,28)(H,31,32)(H3,20,22,25,26,30)/t12-/m0/s1

InChI key

OVBPIULPVIDEAO-LBPRGKRZSA-N

General description

Folic acid (FA)/pterylmonoglutamic acid helps to block neural tube defects (NTDs).

Application

Folic acid has been used:
  • to study the role of vitamins and amino acids on hybridoma growth and monoclonal antibody production
  • in cells and treatments
  • in perforin gene targeting using the CRISPR/Cas gene editing approach

Biochem/physiol Actions

A nutritional delivery form of folate. Folic acid and its derivatives are essential mediators of one-carbon metabolism within cells.


Still not finding the right product?

Explore all of our products under Folic acid


Storage Class

11 - Combustible Solids

wgk

WGK 1

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library