Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Conjugate:
FITC conjugate
Clone:
polyclonal
Application:
IF (d)
Citations:
15
biological source
goat
Quality Segment
conjugate
FITC conjugate
antibody form
affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
form
buffered aqueous solution
technique(s)
direct immunofluorescence: 1:64
storage temp.
2-8°C
target post-translational modification
unmodified
General description
IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections. Anti-Mouse IgG (γ-chain specific)-FITC antibody is specific for is specific for mouse IgG when tested against purified mouse IgA, IgG (all subclasses), and mouse IgM. Goat anti-mouse IgG is isolated by affinity isolation and conjugated to Fluorescein Isothiocyanate (FITC), isomer I.
Immunogen
Purified mouse IgG
Application
Anti-Mouse IgG (γ-chain specific)-FITC antibody may be used for immunofluorescence of mouse spleen cells at a working antibody dilution of 1:64. For ELISA using mice sera, antibody dilution of 1:50 was used. The antibody was also used in anti-nucear antibody detection using mouse sera and for labeling Saimiri brain endothelial cells for exhaustive photon reassignment microscopy
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
Immunofluorescence (1 paper)
Physical form
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Datasheet
Global Trade Item Number
| SKU | GTIN |
|---|---|
| F8264-2ML | 04061833620472 |
| F8264-1ML | 04061838354549 |