Skip to Content
Merck
All Photos(1)

Key Documents

F5803

Sigma-Aldrich

o-Dianisidine dihydrochloride

Suitable for use in glucose determination

Synonym(s):

3,3′-Dimethoxybenzidine dihydrochloride, Fast Blue B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N2O2 · 2HCl
CAS Number:
Molecular Weight:
317.21
Beilstein:
3917996
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

Quality Level

form

crystalline

pH range

6.95-7.05

mp

268 °C

solubility

water: 2.5 mg/mL, colorless

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

Cl.Cl.COc1cc(ccc1N)-c2ccc(N)c(OC)c2

InChI

1S/C14H16N2O2.2ClH/c1-17-13-7-9(3-5-11(13)15)10-4-6-12(16)14(8-10)18-2;;/h3-8H,15-16H2,1-2H3;2*1H

InChI key

UXTIAFYTYOEQHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A preweighed vial containing 50 mg o-Dianisidine.

Specificity

o-Dianisidine (3,3′-dimethoxybenzidine) is a peroxidase substrate suitable for use in ELISA procedures. This substrate produces a soluble end product that is yellow-orange in color and can be read spectrophotometrically at 405 nm. The reaction may be stopped with 5 M HCl.

Application

o-Dianisidine dihydrochloride is suitable for use in the glucose determination. It is suitable reagent used in the following studies:
  • As substrate for the determination of serum ceruloplasmin from its oxidase activity.[1]
  • Determination of total antioxidant response (TAR) against potent free radical reactions by a novel, colorimetric and fully automated method.[2]
  • As colorimetric indicator for the simultaneous semi-quantitative estimation of glucose, lactate and uric acid on paper-based microfluidic devices (microPAD).[3]
  • Myeloperoxidase activity assay.[4]

Warning

Probably carcinogenic.

enzyme

Product No.
Description
Pricing

standard

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Skin Corr. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wijitar Dungchai et al.
Analytica chimica acta, 674(2), 227-233 (2010-08-04)
We report here the use of multiple indicators for a single analyte for paper-based microfluidic devices (microPAD) in an effort to improve the ability to visually discriminate between analyte concentrations. In existing microPADs, a single dye system is used for
Measurement of ceruloplasmin from its oxidase activity in serum by use of o-dianisidine dihydrochloride.
K H Schosinsky et al.
Clinical chemistry, 20(12), 1556-1563 (1974-12-01)
G B Penner et al.
Journal of dairy science, 92(4), 1725-1733 (2009-03-25)
This study was conducted to determine the effects of feeding Fermenten (Church and Dwight Co., Princeton, NJ) with or without dietary sucrose on ruminal fermentation, apparent total-tract nutrient digestibility, and nutrient utilization. Eight ruminally cannulated Holstein cows (163 +/- 55
G B Penner et al.
Journal of dairy science, 92(6), 2767-2781 (2009-05-19)
The objective of the study was to investigate the fractional rate of volatile fatty acid (VFA) absorption and the expression of genes encoding for transporters and enzymes involved in the absorption and metabolism of VFA in ruminal tissue when cattle
Risheng Ye et al.
The Journal of clinical investigation, 128(3), 1178-1189 (2018-02-20)
The compensatory proliferation of insulin-producing β cells is critical to maintaining glucose homeostasis at the early stage of type 2 diabetes. Failure of β cells to proliferate results in hyperglycemia and insulin dependence in patients. To understand the effect of

Protocols

To standardize a procedure for the enzymatic assay of Diamine Oxidase.

Glucose oxidase activity measured via continuous spectrophotometric assay at 500 nm, indicating glucose oxidation rate.

Glucose oxidase activity measured via continuous spectrophotometric assay at 500 nm, indicating glucose oxidation rate.

Glucose oxidase activity measured via continuous spectrophotometric assay at 500 nm, indicating glucose oxidation rate.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service