Skip to Content
Merck

F5763

Carestream® BioMax® XAR Film

Synonym(s):

autoradiography film, xray film

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41105341
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

size

8 in. (20 cm) × 10 in. (25 cm)

Looking for similar products? Visit Product Comparison Guide

General description

  • Coated with emulsion on both sides of the clear base.
  • Recommended for chemiluminescence.
  • Very versatile for autoradiography with high or low energy isotopes.
  • Sensitivity can be increased with intensifying screens when using low-energy isotopes.
  • When handling film in a darkroom, Kodak LED Safelight Darkroom Illuminator is recommended.
  • All the XAR films are the same, just packaged differently.
Each sheet of film is sealed in a light-tight envelope. With high-energy emitters, the film may be exposed directly through the envelope without the use of exposure cassettes or holders.

Other Notes

Kodak X-OMAT AR film was reformulated to become BioMax XAR film.

Legal Information

BioMax is a registered trademark of Carestream Health, Inc.
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pietro Fratta et al.
The EMBO journal, 37(11) (2018-05-17)
TDP-43 (encoded by the gene TARDBP) is an RNA binding protein central to the pathogenesis of amyotrophic lateral sclerosis (ALS). However, how TARDBP mutations trigger pathogenesis remains unknown. Here, we use novel mouse mutants carrying point mutations in endogenous Tardbp
Gabriela Andrejeva et al.
Autophagy, 16(6), 1044-1060 (2019-09-14)
Macroautophagy/autophagy can enable cancer cells to withstand cellular stress and maintain bioenergetic homeostasis by sequestering cellular components into newly formed double-membrane vesicles destined for lysosomal degradation, potentially affecting the efficacy of anti-cancer treatments. Using 13C-labeled choline and 13C-magnetic resonance spectroscopy
Eri Takeda et al.
PloS one, 6(11), e27815-e27815 (2011-11-24)
Vpr, an accessory protein of human immunodeficiency virus type 1, is a multifunctional protein that plays an important role in viral replication. We have previously shown that the region between residues 17 and 74 of Vpr (Vpr(N17C74)) contained a bona

Related Content

Product Information Sheet

Global Trade Item Number

SKUGTIN
F5763-50EA04061834219200

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service